Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01220862_m1
          See other SDCBP GE Assays ›
          Gene Symbol:
          Sdcbp
          Gene Name:
          syndecan binding protein
          Gene Aliases:
          MDA-9, Sycl, syntenin-1
          Chromosome Location:
          Chr.4: 6365680 - 6396122 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01220862_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sdcbp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001098227.1 NP_001091697.1 1 - 2 74 112
          NM_016807.2 NP_058087.2 1 - 2 74 112
          GenBank mRNA AK014678.1 - 48 112
          AK028429.1 - 1 - 2 112
          AK033412.1 - 1 - 2 72 112
          AK146085.1 - 1 - 2 46 112
          AK150278.1 - 1 - 2 39 112
          AK150321.1 - 1 - 2 42 112
          AK150544.1 - 1 - 2 46 112
          AK150623.1 - 1 - 2 50 112
          AK150661.1 - 1 - 2 42 112
          AK150707.1 - 1 - 2 45 112
          AK150721.1 - 1 - 2 54 112
          AK150736.1 - 1 - 2 42 112
          AK150801.1 - 1 - 2 44 112
          AK150996.1 - 1 - 2 46 112
          AK151050.1 - 1 - 2 46 112
          AK152001.1 - 1 - 2 45 112
          AK152017.1 - 1 - 2 46 112
          AK152080.1 - 1 - 2 53 112
          AK152109.1 - 1 - 2 46 112
          AK152434.1 - 1 - 2 46 112
          AK152552.1 - 1 - 2 50 112
          AK152577.1 - 1 - 2 45 112
          AK152855.1 - 1 - 2 46 112
          AK153042.1 - 1 - 2 46 112
          AK153112.1 - 1 - 2 50 112
          AK159124.1 - 1 - 2 42 112
          AK159538.1 - 1 - 2 47 112
          AK159677.1 - 1 - 2 53 112
          AK165657.1 - 1 - 2 44 112
          AK167513.1 - 1 - 2 42 112
          AK168004.1 - 1 - 2 49 112
          AK168777.1 - 1 - 2 42 112
          AK169219.1 - 1 - 2 51 112
          AK169415.1 - 1 - 2 80 112
          BC019400.1 - 1 - 2 4017442 111
          BY779386.1 - 1 - 2 75 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000039.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK028429.1, AK033412.1, AK146085.1, AK150278.1, AK150321.1, AK150544.1, AK150623.1, AK150661.1, AK150707.1, AK150721.1, AK150736.1, AK150801.1, AK150996.1, AK151050.1, AK152001.1, AK152017.1, AK152109.1, AK152434.1, AK152552.1, AK152577.1, AK152855.1, AK153042.1, AK153112.1, AK159538.1, AK159677.1, AK165657.1, AK167513.1, AK168004.1, AK168777.1, AK169219.1, AK169415.1, BC019400.1, NM_001098227.1, NM_016807.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sdcbp

          Gene Ontology Categories:

          Function(s) Process(es)

          Ras protein signal transduction
          positive regulation of phosphorylation
          positive regulation of exosomal secretion
          positive regulation of extracellular exosome assembly
          frizzled binding
          interleukin-5 receptor binding
          protein binding
          protein C-terminus binding
          growth factor binding
          neurexin family protein binding
          identical protein binding
          protein homodimerization activity
          syndecan binding
          ephrin receptor binding
          protein heterodimerization activity
          protein N-terminus binding
          cell adhesion molecule binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400000253.mm.1 TC0400000039.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000253.mm.1 NM_001098227.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000253.mm.1 NM_001098227.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000253.mm.1 NM_016807.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000253.mm.1 NM_016807.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000258.mm.1 TC0400000039.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000258.mm.1 NM_001098227.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000258.mm.1 NM_001098227.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000258.mm.1 NM_016807.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000258.mm.1 NM_016807.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000263.mm.1 TC0400000039.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000263.mm.1 NM_001098227.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000263.mm.1 NM_001098227.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000263.mm.1 NM_016807.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000263.mm.1 NM_016807.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 376402 - -
          ABI Mouse Genome Survey Microarray 740368 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0