Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01221975_g1
          See other PSMA7 GE Assays ›
          Gene Symbol:
          Psma7
          Gene Name:
          proteasome (prosome, macropain) subunit, alpha type 7
          Gene Aliases:
          C6-I
          Chromosome Location:
          Chr.2: 180036366 - 180042464 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01221975_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psma7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289476.1 NP_001276405.1 5 - 6 1322 107
          NM_011969.2 NP_036099.1 5 - 6 683 107
          GenBank mRNA AF019662.1 - 5 - 6 621 107
          AK088615.1 - 5 - 6 628 107
          AK146765.1 - 5 - 6 640 107
          AK165563.1 - 5 - 6 636 107
          AK168234.1 - 5 - 6 654 107
          BC008222.1 - 5 - 6 616 2581897 107
          BC014752.1 - 5 - 6 1270 107
          DV049235.1 - 5 - 6 652 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005399.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psma7

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          ubiquitin-dependent protein catabolic process
          proteasomal ubiquitin-independent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          proteolysis involved in cellular protein catabolic process
          endopeptidase activity
          threonine-type endopeptidase activity
          peptidase activity
          hydrolase activity
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21901047 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21901045 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21901046 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21901044 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21901048 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233603 BC014752.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233603 BC008222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233603 NM_011969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233603 AF019662.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233603 AK088615.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233603 BC014752.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233603 BC008222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233603 NM_011969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233603 AF019662.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233603 AK088615.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488513 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914100 AK008832.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914100 AK030337.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914100 BC036156.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914100 BC036160.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914100 BC100511.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914100 BC117023.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914100 BC117027.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914100 NM_001099917.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18064 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134239 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233603 BC014752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233603 BC008222.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233603 NM_011969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233603 AF019662.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233603 AK088615.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233603 BC014752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233603 BC008222.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233603 NM_011969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233603 AF019662.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233603 AK088615.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P233603 BC014752.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P233603 BC008222.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P233603 NM_011969.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P233603 AF019662.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P233603 AK088615.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P233603 BC014752.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P233603 BC008222.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P233603 NM_011969.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P233603 AF019662.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P233603 AK088615.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21901037 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04807963 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134239 - -
          Whole Mouse Genome G4122A A_51_P233603 BC014752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233603 BC008222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233603 NM_011969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233603 AF019662.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233603 AK088615.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233603 BC014752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233603 BC008222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233603 NM_011969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233603 AF019662.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233603 AK088615.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112480 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01509075 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01509036 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21901045 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21901044 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21901047 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21901046 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21901048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21900944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21901008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01481622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01481621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01631264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01631265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21901027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01479765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01917019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01962605 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10490376 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490377 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490371 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490373 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490372 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490375 - -
          Affymetrix Mouse Gene 1.0 ST Array 10490374 - -
          Affymetrix HT MG-430 PM Array Plate 1423567_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1423568_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10490370 - -
          Mouse Genome 430 2.0 Array 1423567_a_at - -
          Mouse Genome 430 2.0 Array 1423568_PM_at - -
          Mouse Genome 430 2.0 Array 1423567_PM_a_at - -
          Mouse Genome 430 2.0 Array 1423568_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26444_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26444_at - -
          Affymetrix Mouse Clariom S Array PSR0200045874.mm.1 TC0200005399.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045874.mm.1 NM_001289476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045874.mm.1 NM_011969.2 Refseq
          Mouse Expression Set 430 1423568_PM_at - -
          Mouse Expression Set 430 1423568_at - -
          Mouse Expression Set 430 1423567_PM_a_at - -
          Mouse Expression Set 430 1423567_a_at - -
          Murine Genome U74v2 Set 93988_at - -
          Murine Genome U74v2 Set 93988_PM_at - -
          Mouse Genome 430A 2.0 Array 1423567_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1423568_PM_at - -
          Mouse Genome 430A 2.0 Array 1423568_at - -
          Mouse Genome 430A 2.0 Array 1423567_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423567_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1423568_at - -
          Affymetrix HT Mouse Genome 430A Array 1423568_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423567_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2230131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2630328 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2630328 - -
          mouse-6 v1.1 (Illumina) scl18232.3.7_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 2230131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18232.3.7_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2630328 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2793488 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2793488 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2630328 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl18232.3.7_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0