Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01222194_m1
          See other GM5108 GE Assays ›
          Gene Symbol:
          Gm5108
          Gene Name:
          predicted gene 5108
          Gene Aliases:
          EG330097
          Chromosome Location:
          Chr.5: 67941669 - 67977070 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01222194_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5108
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001256184.1 NP_001243113.1 3 - 4 317 78
          GenBank mRNA AK076825.1 - 3 - 4 319 78
          BC120772.1 - 3 - 4 247 78
          BC120798.1 - 3 - 4 247 78
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB615379.1 - 3 - 4 319 Chr. 5 - 67941669 - 67977072 [+] on Build GRCm38 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4930425K10 gene 4930425K10Rik -32.1kb Anti-sense Chr. 5 - 67933612 - 67943081 on NCBI Build GRCm38
          Downstream glutaredoxin,
          cysteine rich 1
          Grxcr1 Tg(Eno2-Gabrb3)0370Brll,
          pi,
          tde
          54.8kb Sense Chr. 5 - 68031835 - 68166398 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000656.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P428801 BC120772.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P428801 BC120798.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P428801 AK076825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P428801 BC120772.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P428801 BC120798.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P428801 AK076825.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855207 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464305 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499545 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805115 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653723 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122504 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136652 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P428801 BC120772.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P428801 BC120798.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P428801 AK076825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P428801 BC120772.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P428801 BC120798.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P428801 AK076825.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01124924 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136652 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05603488 - -
          Whole Mouse Genome G4122A A_52_P428801 BC120772.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P428801 BC120798.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P428801 AK076825.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P428801 BC120772.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P428801 BC120798.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P428801 AK076825.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04327720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23771042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04327718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04107655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04483565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23771046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23771039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04343333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04941630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04395482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23771051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23771048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23771041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04190452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23771049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04632849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04395469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04343337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04107649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04982228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23770812 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500005824.mm.1 TC0500000656.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500005824.mm.1 NM_001256184.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500005824.mm.1 NM_001256184.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500005822.mm.1 TC0500000656.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500005822.mm.1 NM_001256184.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500005822.mm.1 NM_001256184.1 Refseq
          Murine Genome U74v2 Set 170615_PM_at - -
          Murine Genome U74v2 Set 170615_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1250447 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250447 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0