Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01222737_m1
          See other 4930431F12RIK GE Assays ›
          Gene Symbol:
          4930431F12Rik
          Gene Name:
          RIKEN cDNA 4930431F12 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 44966201 - 44978805 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01222737_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930431F12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_073013.1 9 - 10 1271 145
          GenBank mRNA AK015267.1 - 9 - 10 1272 145

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream LIM domain binding 2 Ldb2 AI035351,
          AW146358,
          CLIM1,
          CLP-36,
          Ldb3
          -177.3kb Anti-sense Chr. 5 - 44472131 - 44799760 on NCBI Build GRCm38
          Downstream DNA segment,
          Chr 5,
          ERATO Doi 615,
          expressed
          D5Ertd615e 4930435H24Rik 131.1kb Anti-sense Chr. 5 - 45109944 - 45295343 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000483.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P220543 AK015267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P220543 AK015267.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175861 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116260 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P220543 AK015267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P220543 AK015267.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175861 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116260 - -
          Whole Mouse Genome G4122A A_52_P252034 AK015267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220543 AK015267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220543 AK015267.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125195 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116260 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04170227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654397 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04170224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23654399 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454029_PM_at - -
          Mouse Genome 430 2.0 Array 1454029_at - -
          Mouse Genome 430 2.0 Array 1454029_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500004569.mm.1 TC0500000483.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500004569.mm.1 NR_073013.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004570.mm.1 TC0500000483.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500004570.mm.1 NR_073013.1 Refseq
          Mouse Expression Set 430 1454029_PM_at - -
          Mouse Expression Set 430 1454029_at - -
          Affymetrix HT Mouse Genome 430B Array 1454029_at - -
          Affymetrix HT Mouse Genome 430B Array 1454029_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 336688 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-fb85479f-kf8ql:80/100.66.130.16:80.
          git-commit: 1c69677c60948f80141da00d3c5091e873d99202
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline