Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01222894_gH
          See other DKC1 GE Assays ›
          Gene Symbol:
          Dkc1
          Gene Name:
          dyskeratosis congenita 1, dyskerin
          Gene Aliases:
          BC068171
          Chromosome Location:
          Chr.X: 75095854 - 75109777 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01222894_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dkc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001030307.2 NP_001025478.1 4 - 5 518 86
          GenBank mRNA AK136418.1 - 4 - 5 303 86
          AK144139.1 - 4 - 5 306 86
          AK166105.1 - 4 - 5 294 86
          AK167973.1 - 4 - 5 269 86
          BC068171.1 - 4 - 5 274 86
          BC099966.1 - 4 - 5 358 6435181 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000735.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA modification
          box H/ACA RNA 3'-end processing
          pseudouridine synthesis
          RNA-dependent DNA biosynthetic process
          rRNA processing
          RNA processing
          cell population proliferation
          RNA modification
          rRNA pseudouridine synthesis
          snRNA pseudouridine synthesis
          positive regulation of telomere maintenance via telomerase
          box H/ACA RNA metabolic process
          ribosome biogenesis
          positive regulation of telomerase activity
          DNA biosynthetic process
          mRNA pseudouridine synthesis
          telomerase activity
          RNA binding
          pseudouridine synthase activity
          isomerase activity
          pivalyl-CoA mutase activity
          o-hydroxylaminobenzoate mutase activity
          lupeol synthase activity
          beta-amyrin synthase activity
          baruol synthase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00005082.mm.1 TC0X00000735.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005082.mm.1 NM_001030307.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005082.mm.1 NM_001030307.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005084.mm.1 TC0X00000735.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005084.mm.1 NM_001030307.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005084.mm.1 NM_001030307.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005081.mm.1 TC0X00000735.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005081.mm.1 NM_001030307.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005081.mm.1 NM_001030307.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-tbs5g:80/100.66.134.240:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0