Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01224444_m1
          See other CLN5 GE Assays ›
          Gene Symbol:
          Cln5
          Gene Name:
          ceroid-lipofuscinosis, neuronal 5
          Gene Aliases:
          A730075N08Rik
          Chromosome Location:
          Chr.14: 103070216 - 103077630 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01224444_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cln5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033242.1 NP_001028414.1 3 - 4 646 60
          GenBank mRNA AK032293.1 - 3 - 4 461 60
          AK043247.1 - 3 - 4 570 60
          AK085741.1 - 3 - 4 647 60
          AK144635.1 - 3 - 4 537 60
          BC025487.1 - 3 - 4 503 60
          BC141314.1 - 3 - 4 540 9055926 60
          BC141315.1 - 3 - 4 540 9055927 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001183.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          signal peptide processing
          lysosome organization
          lysosomal lumen acidification
          visual perception
          neurogenesis
          glycosylation
          mannose binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29863554 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29863555 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29863556 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29863559 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320888 BC141315.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320888 BC025487.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320888 BC141314.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320888 AK144635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320888 AK085741.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320888 BC141315.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320888 BC025487.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320888 BC141314.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320888 AK144635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P320888 AK085741.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103385 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735584 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530691 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719732 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 52387 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82534 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170924 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320888 BC141315.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320888 BC025487.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320888 BC141314.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320888 AK144635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320888 AK085741.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320888 BC141315.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320888 BC025487.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320888 BC141314.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320888 AK144635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P320888 AK085741.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P320888 BC141315.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P320888 BC025487.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P320888 BC141314.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P320888 AK144635.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P320888 AK085741.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P320888 BC141315.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P320888 BC025487.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P320888 BC141314.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P320888 AK144635.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P320888 AK085741.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08221434 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170924 - -
          Whole Mouse Genome G4122A A_51_P320888 BC141315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320888 BC025487.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320888 BC141314.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320888 AK144635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320888 AK085741.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320888 BC141315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320888 BC025487.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320888 BC141314.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320888 AK144635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P320888 AK085741.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170924 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29863556 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29863559 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29863554 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29863555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29863617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11498133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11301923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11515607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11498112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11301930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11245904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11651689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11464975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11464978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11651703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29863546 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10416844 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416847 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416846 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416845 - -
          Affymetrix HT MG-430 PM Array Plate 1426886_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10416843 - -
          Mouse Genome 430 2.0 Array 1426886_at - -
          Mouse Genome 430 2.0 Array 1426886_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 211286_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 211286_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400009270.mm.1 TC1400001183.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009270.mm.1 NM_001033242.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009270.mm.1 NM_001033242.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009267.mm.1 TC1400001183.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009267.mm.1 NM_001033242.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009267.mm.1 NM_001033242.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009268.mm.1 TC1400001183.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009268.mm.1 NM_001033242.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009268.mm.1 NM_001033242.1 Refseq
          Mouse Expression Set 430 1426886_PM_at - -
          Mouse Expression Set 430 1426886_at - -
          Mouse Genome 430A 2.0 Array 1426886_at - -
          Mouse Genome 430A 2.0 Array 1426886_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426886_at - -
          Affymetrix HT Mouse Genome 430A Array 1426886_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 926758 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2639290 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2639290 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2639291 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0