Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01224862_m1
          See other HEPH GE Assays ›
          Gene Symbol:
          Heph
          Gene Name:
          hephaestin
          Gene Aliases:
          C130006F04Rik, Cpl, mKIAA0698, sla
          Chromosome Location:
          Chr.X: 96455436 - 96574485 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01224862_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Heph
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159627.1 NP_001153099.1 2 - 3 467 99
          NM_001159628.1 NP_001153100.1 2 - 3 475 99
          NM_010417.2 NP_034547.2 2 - 3 475 99
          NM_181273.4 NP_851790.1 2 - 3 467 99
          GenBank mRNA AF082567.1 - 2 - 3 587 99
          AK032895.1 - 2 - 3 387 99
          AK081330.1 - 2 - 3 443 99
          AK086923.1 - 2 - 3 386 99
          AK129194.1 - 2 - 3 423 99
          BC048237.1 - 2 - 3 315 6474714 99
          BC054442.1 - 2 - 3 315 6474713 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000919.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AF082567.1, AK081330.1, AK086923.1, BC048237.1, BC054442.1, NM_001159627.1, NM_001159628.1, NM_010417.2, NM_181273.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Heph

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          copper ion transport
          iron ion transport
          cellular iron ion homeostasis
          erythrocyte differentiation
          iron ion homeostasis
          ferroxidase activity
          copper ion binding
          ferrous iron binding
          N-ethylmaleimide reductase activity
          oxidoreductase activity
          reduced coenzyme F420 dehydrogenase activity
          sulfur oxygenase reductase activity
          malolactic enzyme activity
          NADPH:sulfur oxidoreductase activity
          metal ion binding
          epoxyqueuosine reductase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00005912.mm.1 TC0X00000919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005912.mm.1 NM_001159627.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005912.mm.1 NM_001159627.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005912.mm.1 NM_181273.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005912.mm.1 NM_181273.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005914.mm.1 TC0X00000919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005914.mm.1 NM_001159627.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005914.mm.1 NM_001159627.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005914.mm.1 NM_001159628.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005914.mm.1 NM_001159628.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005914.mm.1 NM_010417.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005914.mm.1 NM_010417.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005914.mm.1 NM_181273.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005914.mm.1 NM_181273.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005909.mm.1 TC0X00000919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005909.mm.1 NM_001159627.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005909.mm.1 NM_001159627.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005909.mm.1 NM_181273.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005909.mm.1 NM_181273.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005911.mm.1 TC0X00000919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005911.mm.1 NM_001159627.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005911.mm.1 NM_001159627.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005911.mm.1 NM_181273.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005911.mm.1 NM_181273.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 874447 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0