Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01228831_g1
          See other 5530601H04RIK GE Assays ›
          Gene Symbol:
          5530601H04Rik
          Gene Name:
          RIKEN cDNA 5530601H04 gene
          Gene Aliases:
          AI835882
          Chromosome Location:
          Chr.X: 105040854 - 105070124 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01228831_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5530601H04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015467.1 1 - 2 122 66
          GenBank mRNA AK017459.1 - 59 66
          AK043766.1 - 95 66
          AK047017.1 - 95 66
          AK080296.1 - 1 - 2 90 66
          AK084653.1 - 1 - 2 95 66
          AK084980.1 - 1 - 2 74 66
          AK085098.1 - 1 - 2 67 66
          AK085485.1 - 1 - 2 61 66
          AK153718.1 - 41 66
          AK164650.1 - 1 - 2 123 66
          BC055463.1 - 1 - 2 76 66
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB660538.1 - 1 - 2 123 Chr. X - 105043908 - 105070124 [-] on Build GRCm38 66
          GenBankmRNA BB646114.1 - 1 - 2 95 Chr. X - 105039687 - 105070096 [-] on Build GRCm38 66
          GenBankmRNA BU614208.1 - 1 - 2 106 Chr. X - 105039580 - 105070095 [-] on Build GRCm38 66
          GenBankmRNA CF617301.1 - 1 - 2 71 Chr. X - 105043803 - 105070073 [-] on Build GRCm38 66
          GenBankmRNA BG863873.1 - 1 - 2 92 Chr. X - 105067973 - 105070094 [-] on Build GRCm38 66
          GenBankmRNA CF749494.1 - 1 - 2 56 Chr. X - 105042749 - 105070058 [-] on Build GRCm38 66
          GenBankmRNA BY198982.1 - 1 - 2 59 Chr. X - 105066808 - 105070060 [-] on Build GRCm38 66
          GenBankmRNA CA886904.2 - 1 - 2 118 Chr. X - 105047909 - 105070107 [-] on Build GRCm38 66
          GenBankmRNA BB659196.1 - 1 - 2 95 Chr. X - 105044021 - 105070096 [-] on Build GRCm38 66
          GenBankmRNA BG695296.1 - 1 - 2 86 Chr. X - 105043978 - 105070085 [-] on Build GRCm38 66
          GenBankmRNA BB661289.1 - 1 - 2 59 Chr. X - 105043986 - 105070060 [-] on Build GRCm38 66
          GenBankmRNA DV061663.1 - 1 - 2 165 Chr. X - 105043805 - 105070111 [-] on Build GRCm38 66
          GenBankmRNA CK619918.1 - 1 - 2 45 Chr. X - 105044061 - 105070047 [-] on Build GRCm38 66
          GenBankmRNA BY745619.1 - 1 - 2 90 Chr. X - 105044039 - 105070092 [-] on Build GRCm38 66
          GenBankmRNA BB601826.1 - 1 - 2 107 Chr. X - 105068530 - 105070108 [-] on Build GRCm38 66
          GenBankmRNA BB660227.1 - 1 - 2 67 Chr. X - 105044021 - 105070068 [-] on Build GRCm38 66
          GenBankmRNA W11282.1 - 55 Chr. - - [-] on Build 66
          GenBankmRNA BY044552.1 - 1 - 2 77 Chr. X - 105047952 - 105070079 [-] on Build GRCm38 66
          GenBankmRNA BY723805.1 - 1 - 2 109 Chr. X - 105039667 - 105070111 [-] on Build GRCm38 66
          GenBankmRNA BB659892.1 - 1 - 2 74 Chr. X - 105044025 - 105070071 [-] on Build GRCm38 66
          GenBankmRNA BB641536.1 - 1 - 2 95 Chr. X - 105039797 - 105070096 [-] on Build GRCm38 66
          GenBankmRNA BF661519.1 - 1 - 2 57 Chr. X - 105043983 - 105070059 [-] on Build GRCm38 66
          GenBankmRNA BY054683.1 - 1 - 2 77 Chr. X - 105047966 - 105070079 [-] on Build GRCm38 66
          GenBankmRNA BE647238.1 - 1 - 2 55 Chr. X - 105044019 - 105070057 [-] on Build GRCm38 66

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream melanoma antigen,
          family E,
          2
          Magee2 9630059J11Rik,
          BB131182,
          Mage-e2
          -212.3kb Sense Chr. X - 104854952 - 104857267 on NCBI Build GRCm38
          Downstream polysaccharide biosynthesis domain containing 1 Pbdc1 2610029G23Rik 9.7kb Anti-sense Chr. X - 105079756 - 105117094 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 537690 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0