Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01229749_m1
          See other 4933424G06RIK GE Assays ›
          Gene Symbol:
          4933424G06Rik
          Gene Name:
          RIKEN cDNA 4933424G06 gene
          Gene Aliases:
          1700104I15Rik
          Chromosome Location:
          Chr.1: 36714538 - 36757960 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01229749_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933424G06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040290.1 8 - 9 956 73
          GenBank mRNA AK016893.1 - 8 - 9 956 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000254.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760446 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782967 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78388 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380431 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 372905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 357935 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26647 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472892 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 422949 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 72457 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 89005 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136849 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129994 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P397117 AK016893.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P397117 AK016893.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397117 AK016893.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397117 AK016893.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P00270038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20149988 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430235_PM_at - -
          Mouse Genome 430 2.0 Array 1430235_at - -
          Mouse Genome 430 2.0 Array 1430235_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71166_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71166_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100001716.mm.1 TC0100000254.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100001716.mm.1 NR_040290.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001716.mm.1 NR_040290.1 Refseq
          Mouse Expression Set 430 1430235_PM_at - -
          Mouse Expression Set 430 1430235_at - -
          Murine Genome U74v2 Set 168647_PM_at - -
          Murine Genome U74v2 Set 168647_at - -
          Affymetrix HT Mouse Genome 430B Array 1430235_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430235_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 512058 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1240736 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2602314 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2602314 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0