Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01230317_g1
          See other 1700100I10RIK GE Assays ›
          Gene Symbol:
          1700100I10Rik
          Gene Name:
          RIKEN cDNA 1700100I10 gene
          Gene Aliases:
          1700085D13Rik, 4930481K08Rik
          Chromosome Location:
          Chr.14: 114940225 - 114945948 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01230317_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700100I10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015607.1 - 2 - 3 168 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4930524C18 gene 4930524C18Rik -83.7kb Anti-sense Chr. 14 - 114831965 - 114861327 on NCBI Build GRCm38
          Downstream microRNA 17 Mir17 Mirn17,
          mir-17,
          mmu-mir-17
          98.2kb Anti-sense Chr. 14 - 115043671 - 115043754 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 489825 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08244439 - -
          Whole Mouse Genome G4122A A_52_P265785 AK007006.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P265785 AK007101.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P265785 AK015607.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11267723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11692428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11617358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11267720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11731008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11617360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11692433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11294079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11690764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11617372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29918826 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453679_PM_at - -
          Mouse Genome 430 2.0 Array 1453679_PM_at - -
          Mouse Genome 430 2.0 Array 1453679_at - -
          Mouse Expression Set 430 1453679_at - -
          Mouse Expression Set 430 1453679_PM_at - -
          Murine Genome U74v2 Set 168573_PM_at - -
          Murine Genome U74v2 Set 168573_at - -
          Affymetrix HT Mouse Genome 430B Array 1453679_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453679_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc