Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900024431.mm.1 |
TC0900002968.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024431.mm.1 |
NM_027918.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024431.mm.1 |
NM_027918.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024433.mm.1 |
TC0900002968.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024433.mm.1 |
NM_027918.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024433.mm.1 |
NM_027918.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024434.mm.1 |
TC0900002968.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024434.mm.1 |
NM_027918.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024434.mm.1 |
NM_027918.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024435.mm.1 |
TC0900002968.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024435.mm.1 |
NM_027918.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024435.mm.1 |
NM_027918.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024432.mm.1 |
TC0900002968.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900024432.mm.1 |
NM_027918.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900024432.mm.1 |
NM_027918.2 |
Refseq |