Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01230860_s1
          See other HNRNPUL2 GE Assays ›
          Gene Symbol:
          Hnrnpul2
          Gene Name:
          heterogeneous nuclear ribonucleoprotein U-like 2
          Gene Aliases:
          1110031M08Rik, AI465155, Hnrpul2
          Chromosome Location:
          Chr.19: 8819401 - 8834142 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01230860_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hnrnpul2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081196.1 NP_001074665.1 14 5529 146
          GenBank mRNA AK004010.1 - 1 662 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000159.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          RNA binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P18167 NM_001081196.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P18167 AK004010.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464779 AK078249.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 83369 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549490 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897008 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363077 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122484 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P18167 NM_001081196.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P18167 AK004010.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P308828 AK032708.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14502905 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14502904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14502900 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461229 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461226 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461221 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461223 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461236 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461227 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461230 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461228 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461232 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461224 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461231 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461234 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461233 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461235 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461225 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461222 - -
          Affymetrix HT MG-430 PM Array Plate 1434551_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1415765_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461220 - -
          Mouse Genome 430 2.0 Array 1415765_PM_at - -
          Mouse Genome 430 2.0 Array 1434551_PM_at - -
          Mouse Genome 430 2.0 Array 1434551_at - -
          Mouse Genome 430 2.0 Array 1415765_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68693_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68693_at - -
          Affymetrix Mouse Clariom S Array PSR1900002117.mm.1 TC1900000159.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002117.mm.1 NM_001081196.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002117.mm.1 NM_001081196.1 Refseq
          Mouse Expression Set 430 1415765_at - -
          Mouse Expression Set 430 1415765_PM_at - -
          Mouse Expression Set 430 1434551_PM_at - -
          Mouse Expression Set 430 1434551_at - -
          Mouse Expression Set 430 1415765_PM_at - -
          Mouse Expression Set 430 1415765_at - -
          Murine Genome U74v2 Set 104296_at - -
          Murine Genome U74v2 Set 104296_PM_at - -
          Murine Genome U74v2 Set 104609_at - -
          Murine Genome U74v2 Set 104609_PM_at - -
          Mouse Genome 430A 2.0 Array 1434551_at - -
          Mouse Genome 430A 2.0 Array 1415765_PM_at - -
          Mouse Genome 430A 2.0 Array 1415765_at - -
          Mouse Genome 430A 2.0 Array 1434551_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415765_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434551_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434551_at - -
          Affymetrix HT Mouse Genome 430A Array 1415765_at - -
          Affymetrix HT Mouse Genome 430B Array 1415765_at - -
          Affymetrix HT Mouse Genome 430B Array 1415765_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 504730 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106040575 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2666851 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220918 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247431 - -
          Illumina Sentrix mouse-6.v1 bead array 106040575 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106040575 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220918 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0