Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01230900_g1
          See other TMEM179B GE Assays ›
          Gene Symbol:
          Tmem179b
          Gene Name:
          transmembrane protein 179B
          Gene Aliases:
          0610031L02Rik, 1500003K22Rik, 1810059G22Rik
          Chromosome Location:
          Chr.19: 8772522 - 8774467 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01230900_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem179b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026325.3 NP_080601.2 4 - 5 534 74
          NR_028418.1 2 - 3 211 74
          NR_028419.1 4 - 5 464 74
          GenBank mRNA AK002731.1 - 2 - 3 189 74
          AK005135.1 - 4 - 5 453 74
          AK007906.1 - 4 - 5 510 74
          AK010996.1 - 4 - 5 515 74
          BC061068.1 - 4 - 5 434 74
          BC087914.1 - 4 - 5 450 74
          BC116961.1 - 4 - 5 534 40126280 74
          BC116963.1 - 4 - 5 534 40126282 74
          BC144813.1 - 4 - 5 464 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001769.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem179b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31943526 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31943527 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P228031 BC061068.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P228031 BC087914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P228031 BC116961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P228031 BC116963.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P228031 BC144813.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P228031 BC061068.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P228031 BC087914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P228031 BC116961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P228031 BC116963.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P228031 BC144813.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 850449 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P228031 BC061068.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P228031 BC087914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P228031 BC116961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P228031 BC116963.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P228031 BC144813.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P228031 BC061068.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P228031 BC087914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P228031 BC116961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P228031 BC116963.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P228031 BC144813.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31943520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31943526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31943513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31943523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31943519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31943515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31943516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31943521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31943518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31943525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31943522 - -
          Whole Mouse Genome G4122A A_51_P416625 BC061068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416625 BC087914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416625 BC116961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416625 BC116963.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416625 BC144813.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228031 BC061068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228031 BC087914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228031 BC116961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228031 BC116963.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228031 BC144813.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228031 BC061068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228031 BC087914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228031 BC116961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228031 BC116963.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228031 BC144813.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114901 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137028 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14384254 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31943527 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31943526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31943440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31943515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31943525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31943511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14384249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31943513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31943518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31943520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31943522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31943526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31943519 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465788 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465786 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465789 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465791 - -
          Affymetrix HT MG-430 PM Array Plate 1455874_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436963_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1449842_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465790 - -
          Affymetrix Mouse Gene 1.1 ST Array 10465783 - -
          Mouse Genome 430 2.0 Array 1449842_PM_at - -
          Mouse Genome 430 2.0 Array 1455874_PM_at - -
          Mouse Genome 430 2.0 Array 1449842_at - -
          Mouse Genome 430 2.0 Array 1436963_PM_x_at - -
          Mouse Genome 430 2.0 Array 1455874_at - -
          Mouse Genome 430 2.0 Array 1436963_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67706_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67706_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900009978.mm.1 TC1900001769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009978.mm.1 NM_026325.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009978.mm.1 NM_026325.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009978.mm.1 NR_028418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009978.mm.1 NR_028418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009978.mm.1 NR_028419.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009978.mm.1 NR_028419.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009977.mm.1 TC1900001769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009977.mm.1 NM_026325.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009977.mm.1 NM_026325.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009977.mm.1 NR_028418.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009977.mm.1 NR_028418.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009977.mm.1 NR_028419.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009977.mm.1 NR_028419.1 Refseq
          Mouse Expression Set 430 1455874_PM_at - -
          Mouse Expression Set 430 1436963_x_at - -
          Mouse Expression Set 430 1449842_at - -
          Mouse Expression Set 430 1436963_PM_x_at - -
          Mouse Expression Set 430 1455874_at - -
          Mouse Expression Set 430 1449842_PM_at - -
          Murine Genome U74v2 Set 160497_at - -
          Murine Genome U74v2 Set 160497_PM_at - -
          Mouse Genome 430A 2.0 Array 1455874_at - -
          Mouse Genome 430A 2.0 Array 1449842_at - -
          Mouse Genome 430A 2.0 Array 1436963_x_at - -
          Mouse Genome 430A 2.0 Array 1449842_PM_at - -
          Mouse Genome 430A 2.0 Array 1436963_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1455874_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455874_at - -
          Affymetrix HT Mouse Genome 430A Array 1436963_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1436963_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1455874_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449842_at - -
          Affymetrix HT Mouse Genome 430A Array 1449842_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 930156 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6860064 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860064 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2656071 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2656071 - -
          mouse-6 v1.1 (Illumina) scl067706.2_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 6860064 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067706.2_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860064 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2656071 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2656071 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl067706.2_5-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0