Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01231514_g1
          See other BANF1 GE Assays ›
          Gene Symbol:
          Banf1
          Gene Name:
          barrier to autointegration factor 1
          Gene Aliases:
          Baf, Bcrp1, L2bp1/Baf
          Chromosome Location:
          Chr.19: 5364633 - 5366876 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01231514_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Banf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001038231.2 NP_001033320.1 2 - 3 235 97
          NM_001286608.1 NP_001273537.1 2 - 3 577 97
          NM_011793.3 NP_035923.1 2 - 3 443 97
          GenBank mRNA AB025349.1 - 2 - 3 175 97
          AF037080.1 - 1 - 2 118 97
          AK003025.1 - 2 - 3 219 97
          AK004212.1 - 2 - 3 202 97
          AK010257.1 - 2 - 3 221 97
          AK011079.1 - 2 - 3 224 97
          AK011100.1 - 2 - 3 222 97
          AK012588.1 - 2 - 3 205 97
          AK088896.1 - 2 - 3 199 97
          AK168039.1 - 2 - 3 225 96
          BC003709.1 - 2 - 3 212 3599828 97
          CK780850.1 - 2 - 3 207 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000960.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Banf1

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA integration
          establishment of integrated proviral latency
          DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928281 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928285 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928282 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928284 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314019 CK780850.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314019 AK010257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314019 AK011079.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314019 AK011100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314019 AK012588.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314019 CK780850.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314019 AK010257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314019 AK011079.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314019 AK011100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314019 AK012588.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724369 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110968 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314019 CK780850.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314019 AK010257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314019 AK011079.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314019 AK011100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314019 AK012588.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314019 CK780850.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314019 AK010257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314019 AK011079.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314019 AK011100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314019 AK012588.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314019 CK780850.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314019 AK010257.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314019 AK011079.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314019 AK011100.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314019 AK012588.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314019 CK780850.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314019 AK010257.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314019 AK011079.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314019 AK011100.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P314019 AK012588.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928262 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110968 - -
          Whole Mouse Genome G4122A A_51_P314019 CK780850.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314019 AK010257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314019 AK011079.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314019 AK011100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314019 AK012588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314019 CK780850.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314019 AK010257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314019 AK011079.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314019 AK011100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314019 AK012588.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122466 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137715 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120708 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110968 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14334274 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14286745 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14334277 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928285 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928282 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928281 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31928284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14380281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14334277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14450206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14450210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14450207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14401674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14380282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928245 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465008 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465010 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465007 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465009 - -
          Affymetrix HT MG-430 PM Array Plate 1421081_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1421082_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1421083_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465005 - -
          Mouse Genome 430 2.0 Array 1421082_s_at - -
          Mouse Genome 430 2.0 Array 1421083_x_at - -
          Mouse Genome 430 2.0 Array 1421081_PM_a_at - -
          Mouse Genome 430 2.0 Array 1421083_PM_x_at - -
          Mouse Genome 430 2.0 Array 1421081_a_at - -
          Mouse Genome 430 2.0 Array 1421082_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23825_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23825_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900008516.mm.1 TC1900000960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008516.mm.1 NM_001038231.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008516.mm.1 NM_001286608.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008516.mm.1 NM_011793.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008516.mm.1 NR_104482.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008520.mm.1 TC1900000960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008520.mm.1 NM_001286608.1 Refseq
          Mouse Expression Set 430 1421082_s_at - -
          Mouse Expression Set 430 1421083_x_at - -
          Mouse Expression Set 430 1421081_a_at - -
          Mouse Expression Set 430 1421083_PM_x_at - -
          Mouse Expression Set 430 1421081_PM_a_at - -
          Mouse Expression Set 430 1421082_PM_s_at - -
          Murine Genome U74v2 Set 101105_at - -
          Murine Genome U74v2 Set 101105_PM_at - -
          Mouse Genome 430A 2.0 Array 1421083_x_at - -
          Mouse Genome 430A 2.0 Array 1421081_a_at - -
          Mouse Genome 430A 2.0 Array 1421081_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1421083_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1421082_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1421082_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1421082_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1421081_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421083_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1421082_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1421083_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1421081_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 915586 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2850170 - -
          Illumina mouse-6 v1.0 expression beadchip 2850170 - -
          Illumina mouse-6 v1.0 expression beadchip 460164 - -
          Illumina mouse-6 v1.0 expression beadchip 6110139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006110139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000460164 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850170 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2745971 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2698392 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238642 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238642 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2745971 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2698392 - -
          mouse-6 v1.1 (Illumina) scl000538.1_65-S - -
          mouse-6 v1.1 (Illumina) scl023825.3_146-S - -
          mouse-6 v1.1 (Illumina) scl0023825.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 6110139 - -
          Illumina Sentrix mouse-6.v1 bead array 460164 - -
          Illumina Sentrix mouse-6.v1 bead array 2850170 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl023825.3_146-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0023825.1_1-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000538.1_65-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460164 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850170 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2745971 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2685806 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2698392 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3158909 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2685806 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2745971 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2698392 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3158909 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0023825.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000538.1_65-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl023825.3_146-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0