Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01231526_s1
          See other MALAT1 GE Assays ›
          Gene Symbol:
          Malat1
          Gene Name:
          metastasis associated lung adenocarcinoma transcript 1 (non-coding RNA)
          Gene Aliases:
          2210401K01Rik, 9430072K23Rik, AI647968, Neat2
          Chromosome Location:
          Chr.19: 5795690 - 5802671 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01231526_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Malat1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002847.2 1 6806 101
          GenBank mRNA EF177380.1 - 1 6806 101
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB734161.1 - 1 229 Chr. 19 - 5795691 - 5796094 [-] on Build GRCm38 101
          GenBankmRNA BF226057.1 - 1 Chr. 19 - 5795689 - 5796150 [+] on Build GRCm38 101
          GenBankmRNA BE995883.1 - 1 Chr. 19 - 5795694 - 5795997 [+] on Build GRCm38 101
          GenBankmRNA CF982785.1 - 156 Chr. - - [-] on Build 101
          GenBankmRNA BB462054.1 - 1 155 Chr. 19 - 5795690 - 5796020 [-] on Build GRCm38 101
          GenBankmRNA AY722410.1 - 1 6806 Chr. - - [-] on Build 101
          GenBankmRNA BX632354.1 - 1 Chr. 19 - 5795705 - 5795949 [+] on Build GRCm38 101
          GenBankmRNA AA060880.1 - 1 397 Chr. 19 - 5795765 - 5796262 [-] on Build GRCm38 101
          GenBankmRNA CB319789.1 - 2 461 Chr. 19 - 5795704 - 5797036 [-] on Build GRCm38 101
          GenBankmRNA BE687135.1 - 1 Chr. 19 - 5795690 - 5796081 [+] on Build GRCm38 101
          GenBankmRNA BE631854.1 - 1 268 Chr. 19 - 5795690 - 5796133 [-] on Build GRCm38 101
          GenBankmRNA BE631176.1 - 1 Chr. 19 - 5795690 - 5796115 [+] on Build GRCm38 101
          GenBankmRNA AI853644.1 - 1 Chr. 19 - 5795690 - 5796177 [+] on Build GRCm38 101
          GenBankmRNA BY537716.1 - 1 249 Chr. 19 - 5795691 - 5796115 [-] on Build GRCm38 101

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream SCY1-like 1 (S. cerevisiae) Scyl1 2810011O19Rik,
          C85140,
          Ntkl,
          mdf,
          mfd,
          p105
          -24.3kb Sense Chr. 19 - 5758427 - 5771414 on NCBI Build GRCm38
          Downstream nuclear paraspeckle assembly transcript 1 (non-protein coding) Neat1 2310043N10Rik,
          VINC
          39.6kb Sense Chr. 19 - 5842302 - 5845478 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000984.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          nuclear speck organization
          regulation of synapse organization
          positive regulation of myoblast proliferation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021187 EF177380.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021187 NR_002847.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496250 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 932072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968455 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 49937 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74882 - -
          Whole Mouse Genome G4122A A_51_P394383 EF177380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394383 BC004722.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394383 NR_002847.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392598 EF177380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392598 AK020134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P392598 NR_002847.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14284591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31930893 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465245 - -
          Affymetrix HT MG-430 PM Array Plate 1438403_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1452378_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1427285_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1418188_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1436202_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1418189_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1429060_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465244 - -
          Mouse Genome 430 2.0 Array 1438403_PM_s_at - -
          Mouse Genome 430 2.0 Array 1427285_s_at - -
          Mouse Genome 430 2.0 Array 1436202_at - -
          Mouse Genome 430 2.0 Array 1436202_PM_at - -
          Mouse Genome 430 2.0 Array 1452378_PM_at - -
          Mouse Genome 430 2.0 Array 1438403_s_at - -
          Mouse Genome 430 2.0 Array 1427285_PM_s_at - -
          Mouse Genome 430 2.0 Array 1452378_at - -
          Affymetrix Mouse Clariom S Array PSR1900008893.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008893.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008895.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008895.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008891.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008891.mm.1 NR_002847.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900008894.mm.1 TC1900000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900008894.mm.1 NR_002847.3 Refseq
          Mouse Expression Set 430 1438403_s_at - -
          Mouse Expression Set 430 1438403_PM_s_at - -
          Mouse Expression Set 430 1427285_PM_s_at - -
          Mouse Expression Set 430 1452378_PM_at - -
          Mouse Expression Set 430 1427285_s_at - -
          Mouse Expression Set 430 1452378_at - -
          Mouse Expression Set 430 1436202_at - -
          Mouse Expression Set 430 1436202_PM_at - -
          Mouse Genome 430A 2.0 Array 1438403_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1452378_at - -
          Mouse Genome 430A 2.0 Array 1452378_PM_at - -
          Mouse Genome 430A 2.0 Array 1427285_s_at - -
          Mouse Genome 430A 2.0 Array 1438403_s_at - -
          Mouse Genome 430A 2.0 Array 1427285_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1418188_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452378_at - -
          Affymetrix HT Mouse Genome 430A Array 1438403_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452378_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418188_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427285_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1418189_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1438403_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427285_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1418189_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1436202_at - -
          Affymetrix HT Mouse Genome 430B Array 1436202_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429060_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429060_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 713193 - -
          ABI Mouse Genome Survey Microarray 795622 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0