Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01232404_g1
          See other ANAPC13 GE Assays ›
          Gene Symbol:
          Anapc13
          Gene Name:
          anaphase promoting complex subunit 13
          Gene Aliases:
          1810004D07Rik, APC13, SWM1
          Chromosome Location:
          Chr.9: 102626296 - 102634244 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01232404_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Anapc13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181394.3 NP_852059.1 2 - 3 182 65
          GenBank mRNA BC028526.1 - 1 - 2 147 3371464 65
          BY347589.1 - 2 - 3 182 65

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Anapc13

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          cell cycle
          cell division
          protein K11-linked ubiquitination
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781224 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781229 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781232 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781230 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781231 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P368903 NM_181394.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P368903 BC028526.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P368903 NM_181394.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P368903 BC028526.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 607577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361461 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92148 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107305 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P368903 NM_181394.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P368903 BC028526.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P368903 NM_181394.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P368903 BC028526.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P368903 NM_181394.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P368903 BC028526.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P368903 NM_181394.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P368903 BC028526.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26781235 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107305 - -
          Whole Mouse Genome G4122A A_51_P368903 NM_181394.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P368903 BC028526.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P368903 NM_181394.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P368903 BC028526.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129489 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107305 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781231 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781224 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781230 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781232 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26781229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26781230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08043070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08114080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08043073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07500493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07884232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08028853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08028865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08028870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08028856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07521787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07884230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08114084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08043074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07789341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08161273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07500494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26781351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07521793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08028872 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10588225 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588224 - -
          Affymetrix HT MG-430 PM Array Plate 1452586_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10588223 - -
          Mouse Genome 430 2.0 Array 1452586_at - -
          Mouse Genome 430 2.0 Array 1452586_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69010_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69010_PM_at - -
          Mouse Expression Set 430 1452586_at - -
          Mouse Expression Set 430 1452586_PM_at - -
          Murine Genome U74v2 Set 162466_PM_at - -
          Murine Genome U74v2 Set 93539_PM_at - -
          Murine Genome U74v2 Set 162466_at - -
          Murine Genome U74v2 Set 93539_at - -
          Mouse Genome 430A 2.0 Array 1452586_at - -
          Mouse Genome 430A 2.0 Array 1452586_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452586_at - -
          Affymetrix HT Mouse Genome 430A Array 1452586_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3800446 - -
          Illumina mouse-6 v1.0 expression beadchip 3800446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003800446 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220454 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220454 - -
          mouse-6 v1.1 (Illumina) scl069010.2_129-S - -
          Illumina Sentrix mouse-6.v1 bead array 3800446 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl069010.2_129-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3800446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2796089 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2796089 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl069010.2_129-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix