Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01232431_g1
          See other NPHP3 GE Assays ›
          Gene Symbol:
          Nphp3
          Gene Name:
          nephronophthisis 3 (adolescent)
          Gene Aliases:
          3632410F03Rik, AI550417, C230078J01, D330020E01Rik, pcy
          Chromosome Location:
          Chr.9: 104002544 - 104043811 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01232431_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nphp3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028721.3 NP_082997.3 7 - 8 1496 65
          NM_172460.1 NP_766048.1 7 - 8 1496 65
          GenBank mRNA AK048877.1 - 7 - 8 1269 65
          AK052281.1 - 7 - 8 1283 65
          AK133217.1 - 7 - 8 1497 65
          AK173327.1 - 7 - 8 1075 65
          AY259499.1 - 7 - 8 1436 65
          BC115724.1 - 7 - 8 1026 40086844 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003333.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nphp3

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          heart looping
          atrial septum development
          lipid metabolic process
          establishment or maintenance of cell polarity
          determination of left/right symmetry
          Wnt signaling pathway
          extracellular matrix organization
          lung development
          determination of pancreatic left/right asymmetry
          photoreceptor cell maintenance
          maintenance of animal organ identity
          convergent extension involved in gastrulation
          cilium assembly
          kidney morphogenesis
          determination of intestine left/right asymmetry
          determination of stomach left/right asymmetry
          determination of liver left/right asymmetry
          ureter development
          negative regulation of canonical Wnt signaling pathway
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900010201.mm.1 TC0900003333.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900010201.mm.1 NM_028721.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010201.mm.1 NM_028721.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010201.mm.1 NM_172460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010201.mm.1 NM_172460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010202.mm.1 TC0900003333.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900010202.mm.1 NM_028721.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010202.mm.1 NM_028721.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010202.mm.1 NM_172460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010202.mm.1 NM_172460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010203.mm.1 TC0900003333.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900010203.mm.1 NM_028721.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010203.mm.1 NM_028721.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010203.mm.1 NM_172460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010203.mm.1 NM_172460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010205.mm.1 TC0900003333.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900010205.mm.1 NM_028721.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900010205.mm.1 NM_028721.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline