Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01233060_m1
          See other 4732465J04RIK GE Assays ›
          Gene Symbol:
          4732465J04Rik
          Gene Name:
          RIKEN cDNA 4732465J04 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 95773133 - 95794984 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01233060_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4732465J04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK165804.1 - 156 115
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY733809.1 - 1 - 2 156 Chr. 10 - 95778917 - 95795148 [+] on Build GRCm38 115

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream nudix (nucleoside diphosphate linked moiety X)-type motif 4 Nudt4 4933436C10Rik,
          DIPP2,
          DIPP2alpha,
          DIPP2beta,
          HDCMB47P
          -214.8kb Anti-sense Chr. 10 - 95547007 - 95564167 on NCBI Build GRCm38
          Downstream early endosome antigen 1 Eea1 A430109M19Rik,
          B230358H09Rik,
          ZFYVE2
          160.0kb Sense Chr. 10 - 95940660 - 96045518 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P489202 AK169506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P489202 AK165804.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P489202 AK076359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P489202 AK169506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P489202 AK165804.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P489202 AK076359.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881859 AK046778.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881859 BC068167.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881859 BC103671.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881859 BC103672.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881859 BC103673.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881859 BC103674.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881859 D45022.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881859 NM_008426.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 374360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 946219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 221819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 9522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 225423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 958338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211429 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 64206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 122604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74832 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 107104 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133837 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P489202 AK169506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P489202 AK165804.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P489202 AK076359.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P489202 AK169506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P489202 AK165804.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P489202 AK076359.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07159240 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133837 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02194478 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07159164 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07159202 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07159230 - -
          Whole Mouse Genome G4122A A_52_P489202 AK169506.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P489202 AK165804.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P489202 AK076359.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P489202 AK169506.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P489202 AK165804.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P489202 AK076359.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133837 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02194478 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07159164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08313128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08450554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08929983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08825497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08691263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08565206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08678947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08450221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08543136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08577245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08814398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08313131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08334851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08234451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08450550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08577243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08454790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08738918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08335898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08772693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08849461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08497490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08288305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08263054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08432961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08387190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08399452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08772699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08772702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08454794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08387192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08399448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08905389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08334852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27367086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08670103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08772691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27366973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08772695 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10365932 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 103060563 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234752 - -
          Illumina Sentrix mouse-6.v1 bead array 103060563 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103060563 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234752 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0