Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01233093_m1
          See other SIX3OS1 GE Assays ›
          Gene Symbol:
          Six3os1
          Gene Name:
          SIX homeobox 3, opposite strand 1
          Gene Aliases:
          D17Mgi26, E130112H22Rik, Rncr1, Six3os
          Chromosome Location:
          Chr.17: 85601932 - 85618396 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01233093_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Six3os1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038082.1 2 - 3 495 110
          NR_038083.1 2 - 3 495 110
          NR_038085.1 2 - 3 495 110
          NR_038086.1 2 - 3 495 110
          GenBank mRNA AK053722.1 - 2 - 3 497 110
          CA527570.1 - 2 - 3 439 110
          CF737515.1 - 2 - 3 455 110
          CF737925.1 - 2 - 3 471 110
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CV884137.1 - 2 - 3 526 Chr. 17 - 85613681 - 85617637 [-] on Build GRCm38 110
          GenBankmRNA BE332550.1 - 2 - 3 Chr. 17 - 85613784 - 85617436 [+] on Build GRCm38 110
          GenBankmRNA CF729478.1 - 2 - 3 457 Chr. 17 - 85613682 - 85618358 [-] on Build GRCm38 110

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream calmodulin-lysine N-methyltransferase Camkmt 1700106N22Rik,
          AI480743,
          AV099781,
          CLNMT,
          CaM KMT
          -156.1kb Anti-sense Chr. 17 - 85090592 - 85458589 on NCBI Build GRCm38
          Downstream sine oculis-related homeobox 3 Six3 E130112M24Rik,
          Six3a,
          Six3alpha,
          Six3b,
          Six3beta
          5.1kb Anti-sense Chr. 17 - 85620834 - 85626191 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002661.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of photoreceptor cell differentiation
          protein binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145154 AY589791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145154 NR_015387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145164 AY589792.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145164 BC065087.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145164 NR_015385.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145159 AK053722.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145159 BC065087.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145159 AY589790.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145159 NR_015386.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145159 AK021382.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 857857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688015 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 839866 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290076 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 936114 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93746 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163177 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159532 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145164 AY589792.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145164 BC065087.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145164 NR_015385.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145154 AY589791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145154 NR_015387.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145159 AK053722.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145159 BC065087.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145159 AY589790.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145159 NR_015386.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145159 AK021382.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13541037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13541004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13260445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13541054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13302155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13540996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13521393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13541015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13480357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13541000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13302163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13260446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13302159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13540993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13063872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13541001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13541014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13540995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13541049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13540999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31411597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13260447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13511321 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10453392 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453391 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453393 - -
          Affymetrix HT MG-430 PM Array Plate 1431168_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10453390 - -
          Mouse Genome 430 2.0 Array 1431168_PM_at - -
          Mouse Genome 430 2.0 Array 1431168_at - -
          Affymetrix Mouse Clariom S Array PSR1700024202.mm.1 TC1700002661.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700024202.mm.1 NR_038082.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024202.mm.1 NR_038082.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024186.mm.1 TC1700002661.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700024186.mm.1 NR_038085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024186.mm.1 NR_038085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024206.mm.1 TC1700002661.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700024206.mm.1 NR_038082.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024206.mm.1 NR_038082.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024206.mm.1 NR_038083.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024206.mm.1 NR_038083.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024206.mm.1 NR_038084.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024206.mm.1 NR_038084.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024206.mm.1 NR_038085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024206.mm.1 NR_038085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024206.mm.1 NR_038086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024206.mm.1 NR_038086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024187.mm.1 TC1700002661.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700024187.mm.1 NR_038085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024187.mm.1 NR_038085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024205.mm.1 TC1700002661.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700024205.mm.1 NR_038082.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024205.mm.1 NR_038082.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024203.mm.1 TC1700002661.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700024203.mm.1 NR_038082.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024203.mm.1 NR_038082.1 Refseq
          Mouse Expression Set 430 1431168_at - -
          Mouse Expression Set 430 1431168_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431168_at - -
          Affymetrix HT Mouse Genome 430B Array 1431168_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0