Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01233338_s1
          See other 1700020D05RIK GE Assays ›
          Gene Symbol:
          1700020D05Rik
          Gene Name:
          RIKEN cDNA 1700020D05 gene
          Gene Aliases:
          mage-g2
          Chromosome Location:
          Chr.19: 5502767 - 5503787 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01233338_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700020D05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023781.5 NP_076270.3 1 937 74
          GenBank mRNA AK006150.1 - 1 938 74
          BC049585.1 - 1 934 6772543 74
          BC061091.1 - 1 935 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700020D05Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929123 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407866 AK006150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407866 BC049585.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407866 AF319980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407866 BC061091.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407866 NM_023781.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407866 AK006150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407866 BC049585.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407866 AF319980.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407866 BC061091.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407866 NM_023781.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407866 AK006150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407866 BC049585.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407866 AF319980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407866 BC061091.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407866 NM_023781.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407866 AK006150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407866 BC049585.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407866 AF319980.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407866 BC061091.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407866 NM_023781.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407866 AK006150.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407866 BC049585.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407866 AF319980.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407866 BC061091.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407866 NM_023781.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407866 AK006150.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407866 BC049585.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407866 AF319980.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407866 BC061091.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407866 NM_023781.5 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31929129 - -
          Whole Mouse Genome G4122A A_51_P407866 AK006150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407866 BC049585.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407866 AF319980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407866 BC061091.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407866 NM_023781.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407866 AK006150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407866 BC049585.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407866 AF319980.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407866 BC061091.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407866 NM_023781.5 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122029 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14469774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14469781 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14492675 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14290702 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14469779 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929124 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929123 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31929125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31929141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14492697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14450630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14140786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31929127 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465105 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465104 - -
          Affymetrix HT MG-430 PM Array Plate 1422633_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465103 - -
          Mouse Genome 430 2.0 Array 1422633_PM_at - -
          Mouse Genome 430 2.0 Array 1422633_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75555_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75555_PM_at - -
          Mouse Expression Set 430 1422633_PM_at - -
          Mouse Expression Set 430 1422633_at - -
          Murine Genome U74v2 Set 165564_PM_at - -
          Murine Genome U74v2 Set 165564_at - -
          Mouse Genome 430A 2.0 Array 1422633_at - -
          Mouse Genome 430A 2.0 Array 1422633_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422633_at - -
          Affymetrix HT Mouse Genome 430A Array 1422633_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 351073 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2970239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002970239 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2606556 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2606556 - -
          mouse-6 v1.1 (Illumina) scl53499.1.4_164-S - -
          Illumina Sentrix mouse-6.v1 bead array 2970239 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53499.1.4_164-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2970239 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2606555 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2606555 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53499.1.4_164-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0