Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01234280_m1
          See other 4921524L21RIK GE Assays ›
          Gene Symbol:
          4921524L21Rik
          Gene Name:
          RIKEN cDNA 4921524L21 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 6603633 - 6640435 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01234280_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4921524L21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027598.1 NP_081874.1 9 - 10 1112 92
          GenBank mRNA AK014961.1 - 9 - 10 1113 92
          BC049627.1 - 7 - 8 957 6706381 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000046.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986586 AK161257.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986586 BC049627.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986586 NM_027598.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986586 AK014961.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810695 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158545 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135321 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140113 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986586 AK161257.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986586 BC049627.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986586 NM_027598.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986586 AK014961.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158545 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08902968 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135321 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08902913 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140113 - -
          Whole Mouse Genome G4122A A_51_P237815 AK161257.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237815 BC049627.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237815 NM_027598.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P237815 AK014961.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135321 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158545 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31473998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31473996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14115815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31473993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14015550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14079014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13723185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14015552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31473992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31474032 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10453700 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453704 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453695 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453703 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453702 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453696 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453701 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453697 - -
          Affymetrix HT MG-430 PM Array Plate 1431479_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10453694 - -
          Mouse Genome 430 2.0 Array 1431479_at - -
          Mouse Genome 430 2.0 Array 1431479_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70901_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70901_at - -
          Affymetrix Mouse Clariom S Array PSR1800000288.mm.1 TC1800000046.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800000288.mm.1 NM_027598.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800000288.mm.1 NM_027598.1 Refseq
          Mouse Expression Set 430 1431479_PM_at - -
          Mouse Expression Set 430 1431479_at - -
          Affymetrix HT Mouse Genome 430B Array 1431479_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431479_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 496820 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3710601 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2644412 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2644412 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0