Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01235168_m1
          See other 1700044C05RIK GE Assays ›
          Gene Symbol:
          1700044C05Rik
          Gene Name:
          RIKEN cDNA 1700044C05 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 118365878 - 118398165 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01235168_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700044C05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045624.1 5 - 6 555 103
          GenBank mRNA AK006694.1 - 5 - 6 556 103
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY706686.1 - 5 - 6 557 Chr. 14 - 118365878 - 118398170 [+] on Build GRCm38 103

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 9376 Gm9376 EG668814 -126.8kb Sense Chr. 14 - 118267158 - 118267779 on NCBI Build GRCm38
          Downstream ATP-binding cassette,
          sub-family C (CFTR/MRP),
          member 4
          Abcc4 D630049P08Rik,
          MOATB,
          MRP4
          84.5kb Anti-sense Chr. 14 - 118482692 - 118707620 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001283.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 381017 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491275 AK045124.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491275 AK053892.1 genbank_mrna_mouse
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11451941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11786334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11521933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11403390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11825375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11451939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938431 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11833788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11786341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11202216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11823096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11577402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11202215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11833787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29938474 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400009693.mm.1 TC1400001283.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400009693.mm.1 NR_045624.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400009693.mm.1 NR_045624.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 510775 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0