Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01235237_m1
          See other A230004M16RIK GE Assays ›
          Gene Symbol:
          A230004M16Rik
          Gene Name:
          RIKEN cDNA A230004M16 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01235237_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A230004M16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK049150.1 - 4 - 5 534 99
          AK049168.1 - 7 - 8 835 99

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream gamma-aminobutyric acid (GABA) A receptor,
          subunit gamma 2
          Gabrg2 GABAA-R,
          Gabrg-2,
          gamma2
          Anti-sense Chr. 11 - 41910190 - 42000840 on NCBI Build GRCm38
          Downstream gamma-aminobutyric acid (GABA) A receptor,
          subunit alpha 1
          Gabra1 Gabra-1 167.7kb Anti-sense Chr. 11 - 42130939 - 42182951 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2341120 AK038406.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163158 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921427 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 441916 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 408667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262030 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 433274 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 811109 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885365 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785674 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572502 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878760 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 474329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 605501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35754 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 161466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24899 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187607 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264362 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911187 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831723 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 711688 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330303 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 441487 AK079033.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201429 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 680685 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 131213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82112 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 317110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477993 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883689 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342174 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 11818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 516696 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626361 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 588863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190546 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282899 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576817 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 164473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 164615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150923 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 7183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 124899 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149549 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2341120 AK038406.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07318070 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07318013 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07317856 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07318038 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07317975 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07318169 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07317987 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07317919 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02307897 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149549 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07317949 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02308062 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07318082 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07317871 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07318141 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07317830 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07318130 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07317806 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120840 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07317987 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07317871 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07317949 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07318070 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149549 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07317830 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02308062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09263611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09133671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09492163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09122304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08989065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09177195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09702754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09264725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09059848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09718214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09492164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09457288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09457287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27741356 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0