Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01235251_g1
          See other CEBPZOS GE Assays ›
          Gene Symbol:
          Cebpzos
          Gene Name:
          CCAAT/enhancer binding protein (C/EBP), zeta, opposite strand
          Gene Aliases:
          1110001A16Rik
          Chromosome Location:
          Chr.17: 78916499 - 78921048 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01235251_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cebpzos
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177402.1 NP_001170873.1 3 - 4 180 152
          NM_001177403.1 NP_001170874.1 3 - 4 348 152
          GenBank mRNA AK003199.1 - 3 - 4 178 152
          AK027910.1 - 3 - 4 176 152
          AV491230.1 - 3 - 4 329 152

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P222593 NM_001177403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P222593 AK003199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P222593 AK027910.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P222593 NM_001177402.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P222593 AK017537.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P222593 NM_001177403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P222593 AK003199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P222593 AK027910.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P222593 NM_001177402.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P222593 AK017537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P222593 NM_001177403.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P222593 AK003199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P222593 AK027910.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P222593 NM_001177402.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P222593 AK017537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P222593 NM_001177403.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P222593 AK003199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P222593 AK027910.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P222593 NM_001177402.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P222593 AK017537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P222593 NM_001177403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P222593 AK003199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P222593 AK027910.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P222593 NM_001177402.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P222593 AK017537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P222593 NM_001177403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P222593 AK003199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P222593 AK027910.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P222593 NM_001177402.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P222593 AK017537.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13098794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13098799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31374352 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1435755_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10447038 - -
          Mouse Genome 430 2.0 Array 1435755_PM_at - -
          Mouse Genome 430 2.0 Array 1435755_at - -
          Mouse Expression Set 430 1435755_PM_at - -
          Mouse Expression Set 430 1435755_at - -
          Murine Genome U74v2 Set 160709_PM_at - -
          Murine Genome U74v2 Set 160709_at - -
          Mouse Genome 430A 2.0 Array 1435755_at - -
          Mouse Genome 430A 2.0 Array 1435755_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435755_at - -
          Affymetrix HT Mouse Genome 430A Array 1435755_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 902878 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105130494 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix