Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01235254_m1
          See other 4930405O22RIK GE Assays ›
          Gene Symbol:
          4930405O22Rik
          Gene Name:
          RIKEN cDNA 4930405O22 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 63305446 - 63312973 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01235254_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930405O22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK029589.1 - 2 - 3 376 80
          AK160416.1 - 2 - 3 363 80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB614916.1 - 2 - 3 373 Chr. 17 - 63305863 - 63312971 [-] on Build GRCm38 80
          GenBankmRNA AV259712.2 - 1 - 2 224 Chr. 17 - 63305683 - 63308708 [-] on Build GRCm38 80
          GenBankmRNA CR517497.1 - 1 - 2 Chr. 17 - 63305987 - 63308614 [+] on Build GRCm38 80
          GenBankmRNA BY706841.1 - 2 - 3 357 Chr. 17 - 63305823 - 63312955 [-] on Build GRCm38 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream F-box and leucine-rich repeat protein 17 Fbxl17 6330576B01Rik,
          AI452053,
          BB073797,
          C130023C01Rik,
          Fbx13,
          Fbxo13
          Sense Chr. 17 - 63045949 - 63504042 on NCBI Build GRCm38
          Downstream fer (fms/fps related) protein kinase Fer AV082135,
          C330004K01Rik,
          Fert,
          Fert2
          521.2kb Anti-sense Chr. 17 - 63829950 - 64146519 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13098471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13440061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13361175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31289579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13480854 - -
          Array Name Probe IDs Targets Target Type
          Murine Genome U74v2 Set 171246_at - -
          Murine Genome U74v2 Set 171246_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0