Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01236130_g1
          See other PRDX5 GE Assays ›
          Gene Symbol:
          Prdx5
          Gene Name:
          peroxiredoxin 5
          Gene Aliases:
          AOEB166, AOPP, Pmp20, Prdx6, PrxV
          Chromosome Location:
          Chr.19: 6906819 - 6909645 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01236130_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prdx5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012021.2 NP_036151.1 5 - 6 560 68
          GenBank mRNA AF110733.1 - 5 - 6 486 68
          AF124994.1 - 5 - 6 649 68
          AF197951.1 - 5 - 6 560 68
          AK002383.1 - 5 - 6 526 68
          AK003332.2 - 5 - 6 526 68
          AK009858.1 - 5 - 6 995 68
          AK136130.1 - 5 - 6 501 68
          AK152653.1 - 5 - 6 526 68
          BC008174.1 - 5 - 6 945 3481713 68
          BC014682.1 - 3 - 4 170 68

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prdx5

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          response to oxidative stress
          negative regulation of transcription by RNA polymerase III
          positive regulation of collagen biosynthetic process
          cellular response to reactive oxygen species
          hydrogen peroxide catabolic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          negative regulation of oxidoreductase activity
          regulation of apoptosis involved in tissue homeostasis
          NADPH oxidation
          reactive nitrogen species metabolic process
          RNA polymerase III transcription regulatory region sequence-specific DNA binding
          peroxidase activity
          signaling receptor binding
          antioxidant activity
          oxidoreductase activity
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          protein dimerization activity
          peroxiredoxin activity
          peroxynitrite reductase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936608 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936611 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936609 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936610 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979463 BC008174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979463 BC014682.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979463 AF124994.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979463 AF197951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979463 AK002383.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033255 BC008174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033255 BC014682.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033255 AF124994.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033255 AK002383.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2033255 AK009858.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 308125 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305196 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979463 BC008174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979463 BC014682.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979463 AF124994.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979463 AF197951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979463 AK002383.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033255 BC008174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033255 BC014682.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033255 AF124994.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033255 AK002383.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2033255 AK009858.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174591 BC008174.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174591 BC014682.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174591 AF124994.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174591 AF197951.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174591 AK002383.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31936592 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03545335 - -
          Whole Mouse Genome G4122A A_51_P174591 BC008174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174591 BC014682.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174591 AF124994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174591 AF197951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174591 AK002383.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136783 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117648 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14248025 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14248000 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14248008 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14247993 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936608 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936610 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936609 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31936611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31936608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14207529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14485433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14485438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14248025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14234953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14485435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14162600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14255637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14348594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14162596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14348595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14485434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31936590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14247993 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461013 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461014 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465479 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465483 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461015 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465484 - -
          Affymetrix HT MG-430 PM Array Plate 1416381_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465474 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461012 - -
          Mouse Genome 430 2.0 Array 1416381_PM_a_at - -
          Mouse Genome 430 2.0 Array 1416381_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54683_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54683_at - -
          Mouse Expression Set 430 1416381_PM_a_at - -
          Mouse Expression Set 430 1416381_a_at - -
          Murine Genome U74v2 Set 96956_PM_at - -
          Murine Genome U74v2 Set 96956_at - -
          Mouse Genome 430A 2.0 Array 1416381_a_at - -
          Mouse Genome 430A 2.0 Array 1416381_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416381_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416381_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106760035 - -
          Illumina mouse-6 v1.0 expression beadchip 1660592 - -
          Illumina mouse-6 v1.0 expression beadchip 106760035 - -
          Illumina mouse-6 v1.0 expression beadchip 2030091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001660592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002030091 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239765 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2637233 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2637233 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239765 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229380 - -
          mouse-6 v1.1 (Illumina) scl054683.1_165-S - -
          mouse-6 v1.1 (Illumina) scl0054683.2_242-S - -
          mouse-6 v1.1 (Illumina) scl000502.1_59-S - -
          Illumina Sentrix mouse-6.v1 bead array 1660592 - -
          Illumina Sentrix mouse-6.v1 bead array 2030091 - -
          Illumina Sentrix mouse-6.v1 bead array 106760035 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0054683.2_242-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000502.1_59-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660592 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106760035 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2030091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2637233 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1239765 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2637233 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239765 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000502.1_59-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0054683.2_242-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0