Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01236530_g1
          See other ALDH3B2 GE Assays ›
          Gene Symbol:
          Aldh3b2
          Gene Name:
          aldehyde dehydrogenase 3 family, member B2
          Gene Aliases:
          AI848594, C130048D07Rik
          Chromosome Location:
          Chr.19: 3972328 - 3981665 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01236530_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldh3b2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177438.1 NP_001170909.1 3 - 4 562 80
          GenBank mRNA AK136505.1 - 3 - 4 564 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000021.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Process(es)

          molecular_function
          3-chloroallyl aldehyde dehydrogenase activity
          aldehyde dehydrogenase (NAD+) activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900000231.mm.1 TC1900000021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000231.mm.1 NM_001177438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000231.mm.1 NM_001177438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000233.mm.1 TC1900000021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000233.mm.1 NM_001177438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000233.mm.1 NM_001177438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000235.mm.1 TC1900000021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000235.mm.1 NM_001177438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000235.mm.1 NM_001177438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000232.mm.1 TC1900000021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000232.mm.1 NM_001177438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000232.mm.1 NM_001177438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000229.mm.1 TC1900000021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000229.mm.1 NM_001177438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000229.mm.1 NM_001177438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000230.mm.1 TC1900000021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000230.mm.1 NM_001177438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000230.mm.1 NM_001177438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000234.mm.1 TC1900000021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000234.mm.1 NM_001177438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000234.mm.1 NM_001177438.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0