Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01236536_g1
          See other ALDH3B2 GE Assays ›
          Gene Symbol:
          Aldh3b2
          Gene Name:
          aldehyde dehydrogenase 3 family, member B2
          Gene Aliases:
          AI848594, C130048D07Rik
          Chromosome Location:
          Chr.19: 3972328 - 3981665 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01236536_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldh3b2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177438.1 NP_001170909.1 9 - 10 1505 67
          GenBank mRNA AK048310.1 - 366 67
          BC100370.1 - 363 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000021.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Process(es)

          molecular_function
          3-chloroallyl aldehyde dehydrogenase activity
          aldehyde dehydrogenase (NAD+) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108651 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006148 AK048310.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006148 NM_001177438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2168316 NM_001177438.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09089303 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108651 - -
          Whole Mouse Genome G4122A A_52_P116018 AK048310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P116018 BC100370.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P116018 NM_001177438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P116018 AK136505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P442894 AK048310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P442894 AK136505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P442894 NM_001177438.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14330043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920377 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14334766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31920361 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460262 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460260 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460261 - -
          Affymetrix HT MG-430 PM Array Plate 1456317_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460253 - -
          Affymetrix Mouse Gene 1.1 ST Array 10460259 - -
          Mouse Genome 430 2.0 Array 1456317_PM_at - -
          Mouse Genome 430 2.0 Array 1456317_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 621603_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 621603_at - -
          Affymetrix Mouse Clariom S Array PSR1900000238.mm.1 TC1900000021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000238.mm.1 NM_001177438.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000238.mm.1 NM_001177438.1 Refseq
          Mouse Expression Set 430 1456317_PM_at - -
          Mouse Expression Set 430 1456317_at - -
          Murine Genome U74v2 Set 166329_PM_f_at - -
          Murine Genome U74v2 Set 166329_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1456317_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456317_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 326739 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2746937 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2746937 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0