Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700021864.mm.1 |
TC1700002333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700021864.mm.1 |
NM_028782.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021864.mm.1 |
NM_028782.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021867.mm.1 |
TC1700002333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700021867.mm.1 |
NM_028782.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021867.mm.1 |
NM_028782.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021868.mm.1 |
TC1700002333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700021868.mm.1 |
NM_028782.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021868.mm.1 |
NM_028782.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021863.mm.1 |
TC1700002333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700021863.mm.1 |
NM_028782.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021863.mm.1 |
NM_028782.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021865.mm.1 |
TC1700002333.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700021865.mm.1 |
NM_028782.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700021865.mm.1 |
NM_028782.2 |
Refseq |