Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01238934_m1
          See other 4930448I06RIK GE Assays ›
          Gene Symbol:
          4930448I06Rik
          Gene Name:
          RIKEN cDNA 4930448I06 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 41167560 - 41181252 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01238934_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930448I06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040475.1 2 - 3 298 82
          GenBank mRNA AK015419.1 - 2 - 3 299 82
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY714967.1 - 2 - 3 299 Chr. 1 - 41167560 - 41181252 [-] on Build GRCm38 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transmembrane protein 182 Tmem182 2310079P10Rik,
          AI414494
          -310.7kb Anti-sense Chr. 1 - 40805601 - 40856887 on NCBI Build GRCm38
          Downstream POU domain,
          class 3,
          transcription factor 3 adjacent noncoding transcript 1
          Pantr1 2610017I09Rik,
          Pou3f3os,
          linc-Brn1a
          1.5mb Sense Chr. 1 - 42648200 - 42694825 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002254.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC0900001896.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 6365 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 6660 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 92559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174290 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04075384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00152758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00902198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00523115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00152755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00171078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00171075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00902203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00523108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00523099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00902201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00152768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00523121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00152766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00747892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20175978 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100018117.mm.1 TC0100002254.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100018117.mm.1 NR_040475.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018117.mm.1 NR_040475.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018116.mm.1 TC0100002254.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100018116.mm.1 NR_040475.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018116.mm.1 NR_040475.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018115.mm.1 TC0100002254.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100018115.mm.1 NR_040475.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100018115.mm.1 NR_040475.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 490847 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104610154 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0