Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01238949_m1
          See other GM4956 GE Assays ›
          Gene Symbol:
          Gm4956
          Gene Name:
          predicted gene 4956
          Gene Aliases:
          EG241041
          Chromosome Location:
          Chr.1: 21285246 - 21298312 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01238949_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm4956
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002858.1 5 - 6 660 67
          GenBank mRNA BC061226.1 - 5 - 6 660 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream glutathione S-transferase,
          alpha 3
          Gsta3 Gst2-3 -19.7kb Anti-sense Chr. 1 - 21240585 - 21265575 on NCBI Build GRCm38
          Downstream KH domain containing 1A Khdc1a AU014713,
          C88118,
          Ndg1
          41.2kb Anti-sense Chr. 1 - 21329476 - 21352200 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002039.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986341 BC061226.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986341 NR_002858.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133892 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 116241 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 498043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420054 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170843 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117367 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175470 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144570 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144570 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175470 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175470 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00598437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01131533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01131535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00841962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00111163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00593698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00792057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01131540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01057813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00111162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00188051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00694336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00845377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00111159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00792055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00686785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20077222 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10353458 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353457 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353456 - -
          Affymetrix Mouse Gene 1.1 ST Array 10353450 - -
          Affymetrix Mouse Clariom S Array PSR0100016655.mm.1 TC0100002039.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100016655.mm.1 NR_002858.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100016656.mm.1 TC0100002039.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100016656.mm.1 NR_002858.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 689934 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254228 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2911009 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2911009 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-849d5b7584-dvwjr:80/100.66.130.62:80.
          git-commit: 0a9b6045e9144c2c4a21b698be73aa131cdbda4b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0