Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01239081_m1
          See other 4930524O08RIK GE Assays ›
          Gene Symbol:
          4930524O08Rik
          Gene Name:
          RIKEN cDNA 4930524O08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 89826519 - 89864028 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01239081_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930524O08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040735.1 5 - 6 1236 79
          GenBank mRNA AK015891.1 - 5 - 6 1237 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 184 Mir184 Mirn184,
          mir-184,
          mmu-mir-184
          -56.1kb Anti-sense Chr. 9 - 89802260 - 89802328 on NCBI Build GRCm38
          Downstream RAS protein-specific guanine nucleotide-releasing factor 1 Rasgrf1 AI844718,
          CDC25,
          CDC25Mm,
          Gnrp,
          Grf1,
          Grfbeta,
          P190-A,
          Ras-GRF1,
          p190,
          p190RhoGEF
          45.4kb Sense Chr. 9 - 89909472 - 90026979 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001151.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2291621 AK015891.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500463 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417608 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520232 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354363 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351700 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387747 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42571 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27382 - -
          Whole Mouse Genome G4122A A_52_P644091 AK030170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644091 AK015891.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P488531 AK030170.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07958841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07561878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07561880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07958832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07828996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08139475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08165154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07958842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07561884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26714437 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454108_PM_at - -
          Mouse Genome 430 2.0 Array 1454108_PM_at - -
          Mouse Genome 430 2.0 Array 1454108_at - -
          Affymetrix Mouse Clariom S Array PSR0900009017.mm.1 TC0900001151.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900009017.mm.1 NR_040735.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009017.mm.1 NR_040735.1 Refseq
          Mouse Expression Set 430 1454108_PM_at - -
          Mouse Expression Set 430 1454108_at - -
          Affymetrix HT Mouse Genome 430B Array 1454108_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454108_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 509647 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101740139 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix