Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01239760_m1
          See other MRPS22 GE Assays ›
          Gene Symbol:
          Mrps22
          Gene Name:
          mitochondrial ribosomal protein S22
          Gene Aliases:
          3100002P07Rik, Rpms22
          Chromosome Location:
          Chr.9: 98588730 - 98601679 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01239760_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrps22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025485.3 NP_079761.1 7 - 8 1017 83
          GenBank mRNA AK003836.1 - 6 - 7 785 83
          AK013925.1 - 7 - 8 998 83
          AK154600.1 - 7 - 8 998 83
          BC021882.1 - 2 - 3 236 83
          BC051198.1 - 7 - 8 997 1396237 83
          BE983156.1 - 1 - 2 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002908.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrps22

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26759568 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26759566 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362903 AK013925.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362903 AK013925.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824475 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539033 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395129 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 962697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424701 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 8045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52921 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 108187 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178227 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171915 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362903 AK013925.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362903 AK013925.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26759560 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178227 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171915 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02009121 - -
          Whole Mouse Genome G4122A A_51_P362903 AK013925.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362903 AK013925.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125122 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171915 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178227 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07846567 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07846568 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07716463 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08052828 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07596493 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26759568 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26759566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26759520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07846571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08017913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07774565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08052828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08087124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08008491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07596493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07846567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08076272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07473958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08076280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07905072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07846565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07905060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07913195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07905065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26759586 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10595904 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595903 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595905 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595906 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595907 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595908 - -
          Affymetrix Mouse Gene 1.0 ST Array 10595902 - -
          Affymetrix HT MG-430 PM Array Plate 1416595_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10595900 - -
          Mouse Genome 430 2.0 Array 1416595_PM_at - -
          Mouse Genome 430 2.0 Array 1416595_at - -
          Mouse Genome 430 2.0 Array 1443032_at - -
          Mouse Genome 430 2.0 Array 1443032_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 64655_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 64655_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900023953.mm.1 TC0900002908.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900023953.mm.1 NM_025485.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900023953.mm.1 NM_025485.3 Refseq
          Mouse Expression Set 430 1416595_at - -
          Mouse Expression Set 430 1416595_PM_at - -
          Murine Genome U74v2 Set 160621_PM_at - -
          Murine Genome U74v2 Set 160621_at - -
          Mouse Genome 430A 2.0 Array 1416595_at - -
          Mouse Genome 430A 2.0 Array 1416595_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416595_at - -
          Affymetrix HT Mouse Genome 430A Array 1416595_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 855683 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5910458 - -
          Illumina mouse-6 v1.0 expression beadchip 5910458 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005910458 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2771594 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2771594 - -
          mouse-6 v1.1 (Illumina) scl35468.7.1_95-S - -
          Illumina Sentrix mouse-6.v1 bead array 5910458 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35468.7.1_95-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5910458 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2787396 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2771594 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2787396 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0