Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01240547_m1
          See other 4933416E03RIK GE Assays ›
          Gene Symbol:
          4933416E03Rik
          Gene Name:
          RIKEN cDNA 4933416E03 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 159947341 - 159981289 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01240547_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933416E03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040498.1 6 - 7 714 70
          GenBank mRNA AK016829.1 - 6 - 7 716 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream DEAH (Asp-Glu-Ala-His) box polypeptide 35 Dhx35 1200009D07Rik,
          Ddx35
          -1.1mb Sense Chr. 2 - 158794770 - 158858220 on NCBI Build GRCm38
          Downstream RIKEN cDNA 9130015L21 gene 9130015L21Rik 35.4kb Sense Chr. 2 - 160016653 - 160040620 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002446.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2274508 AK016829.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2274508 AK016829.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02280497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02280510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01435329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01907088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02194368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02280501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02194361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01435314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01435321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21791878 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10478193 - -
          Affymetrix HT MG-430 PM Array Plate 1430711_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10478192 - -
          Mouse Genome 430 2.0 Array 1430711_at - -
          Mouse Genome 430 2.0 Array 1430711_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200019915.mm.1 TC0200002446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200019915.mm.1 NR_040498.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200019915.mm.1 NR_040498.1 Refseq
          Mouse Expression Set 430 1430711_PM_at - -
          Mouse Expression Set 430 1430711_at - -
          Murine Genome U74v2 Set 98358_at - -
          Murine Genome U74v2 Set 98358_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430711_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430711_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 886423 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100450168 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix