Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01241416_g1
          See other AKTIP GE Assays ›
          Gene Symbol:
          Aktip
          Gene Name:
          thymoma viral proto-oncogene 1 interacting protein
          Gene Aliases:
          AL023020, Fif, Ft, Ft1, Fts
          Chromosome Location:
          Chr.8: 91109871 - 91135494 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01241416_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aktip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK017861.1 - 5 - 6 577 80
          AK146459.1 - 6 - 7 693 80
          AK158704.1 - 6 - 7 695 80
          BC008130.1 - 7 - 8 698 3498984 80
          BC034861.1 - 6 - 7 702 80
          X71978.1 - 6 - 7 717 80
          Z67963.1 - 6 - 7 717 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002680.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800020489.mm.1 TC0800002680.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800020489.mm.1 NM_010241.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800020490.mm.1 TC0800002680.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800020490.mm.1 NM_010241.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800020491.mm.1 TC0800002680.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800020491.mm.1 NM_010241.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800020497.mm.1 TC0800002680.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800020498.mm.1 TC0800002680.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800020487.mm.1 TC0800002680.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800020487.mm.1 NM_010241.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800020488.mm.1 TC0800002680.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800020488.mm.1 NM_010241.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800020486.mm.1 TC0800002680.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800020486.mm.1 NM_010241.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800020492.mm.1 TC0800002680.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800020492.mm.1 NM_010241.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0