Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01242442_g1
          See other TACO1 GE Assays ›
          Gene Symbol:
          Taco1
          Gene Name:
          translational activator of mitochondrially encoded cytochrome c oxidase I
          Gene Aliases:
          2310066I18Rik, Ccdc44
          Chromosome Location:
          Chr.11: 106066039 - 106073612 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01242442_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Taco1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027346.1 NP_081622.1 4 - 5 828 89
          GenBank mRNA AK047078.1 - 4 - 5 848 89
          AK140143.1 - 4 - 5 866 89
          AK167963.1 - 4 - 5 845 89
          BC029182.1 - 4 - 5 828 4195338 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001771.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Taco1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of translation
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28099379 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28099381 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28099382 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386911 AK140143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386911 AK147942.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386911 AK167963.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386911 BC029182.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386911 NM_027346.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386911 AK140143.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386911 AK147942.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386911 AK167963.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386911 BC029182.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386911 NM_027346.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384061 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166425 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386911 AK140143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386911 AK147942.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386911 AK167963.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386911 BC029182.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386911 NM_027346.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386911 AK140143.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386911 AK147942.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386911 AK167963.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386911 BC029182.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386911 NM_027346.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28099394 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07478699 - -
          Whole Mouse Genome G4122A A_51_P386911 AK140143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386911 AK147942.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386911 AK167963.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386911 BC029182.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386911 NM_027346.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386911 AK140143.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386911 AK147942.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386911 AK167963.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386911 BC029182.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386911 NM_027346.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121001 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116343 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09451064 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09695725 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09695723 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08997766 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28099382 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28099381 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28099379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28099394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09034009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09695720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09060618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09377603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09451063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09365944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09695725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28099369 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382023 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382025 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382027 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382024 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382026 - -
          Affymetrix Mouse Gene 1.1 ST Array 10382022 - -
          Affymetrix Mouse Clariom S Array PSR1100016744.mm.1 TC1100001771.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016744.mm.1 NM_027346.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016744.mm.1 NM_027346.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 367227 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106590484 - -
          Illumina mouse-6 v1.0 expression beadchip 4010129 - -
          Illumina mouse-6 v1.0 expression beadchip 106590484 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004010129 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219565 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221899 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219565 - -
          mouse-6 v1.1 (Illumina) scl40802.5.1_48-S - -
          Illumina Sentrix mouse-6.v1 bead array 4010129 - -
          Illumina Sentrix mouse-6.v1 bead array 106590484 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40802.5.1_48-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106590484 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4010129 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219565 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221899 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219565 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc