Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01242727_m1
          See other ICAM2 GE Assays ›
          Gene Symbol:
          Icam2
          Gene Name:
          intercellular adhesion molecule 2
          Gene Aliases:
          CD102, Icam-2, Ly-60
          Chromosome Location:
          Chr.11: 106377656 - 106388122 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01242727_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Icam2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010494.1 NP_034624.1 3 - 4 805 72
          GenBank mRNA AK007801.1 - 3 - 4 740 72
          BC039128.1 - 3 - 4 737 3962648 72
          BC039970.1 - 3 - 4 757 4487248 72
          X65493.1 - 3 - 4 805 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003923.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Icam2

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          cell-cell adhesion
          integrin binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982049 BC039970.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982049 X65493.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982049 NM_010494.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982049 BC039128.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 833125 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511013 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133794 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125206 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982049 BC039970.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982049 X65493.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982049 NM_010494.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982049 BC039128.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267754 BC039970.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267754 X65493.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267754 NM_010494.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267754 AK007801.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267754 BC039128.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28101326 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125206 - -
          Whole Mouse Genome G4122A A_51_P267754 BC039970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267754 X65493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267754 NM_010494.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267754 AK007801.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267754 BC039128.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138593 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125206 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09146063 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09146074 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09146071 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09146068 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09146058 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09146065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28101307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09385975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09146074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09499604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09499612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09385979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09056820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09146058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09691416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09385977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08994378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09542277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08998801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09146062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09339003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09460692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08994376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09540966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09199698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09578333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09127361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09724637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09146072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09385982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09146068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09199695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09385963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09445211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08994366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28101307 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10392179 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392182 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392181 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392180 - -
          Affymetrix Mouse Gene 1.0 ST Array 10392178 - -
          Affymetrix HT MG-430 PM Array Plate 1448862_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10392177 - -
          Mouse Genome 430 2.0 Array 1448862_PM_at - -
          Mouse Genome 430 2.0 Array 1448862_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15896_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15896_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100036756.mm.1 TC1100003923.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100036756.mm.1 NM_010494.2 Refseq
          Mouse Expression Set 430 1448862_at - -
          Mouse Expression Set 430 1448862_PM_at - -
          Murine Genome U74v2 Set 99053_at - -
          Murine Genome U74v2 Set 99053_PM_at - -
          Mouse Genome 430A 2.0 Array 1448862_PM_at - -
          Mouse Genome 430A 2.0 Array 1448862_at - -
          Affymetrix HT Mouse Genome 430A Array 1448862_at - -
          Affymetrix HT Mouse Genome 430A Array 1448862_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6400113 - -
          Illumina mouse-6 v1.0 expression beadchip 6400113 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006400113 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2618408 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2618408 - -
          mouse-6 v1.1 (Illumina) scl39447.7.1_219-S - -
          Illumina Sentrix mouse-6.v1 bead array 6400113 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39447.7.1_219-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6400113 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2618408 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2618408 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0