Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01243187_g1
          See other P4HB GE Assays ›
          Gene Symbol:
          P4hb
          Gene Name:
          prolyl 4-hydroxylase, beta polypeptide
          Gene Aliases:
          ERp59, PDI, Pdia1, Thbp
          Chromosome Location:
          Chr.11: 120560304 - 120572936 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01243187_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: P4hb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011032.2 NP_035162.1 3 - 4 615 111
          GenBank mRNA AK141304.1 - 3 - 4 552 111
          AK146542.1 - 3 - 4 553 111
          AK149605.1 - 3 - 4 554 111
          AK149965.1 - 3 - 4 553 111
          AK150002.1 - 3 - 4 579 111
          AK150422.1 - 3 - 4 553 111
          AK150592.1 - 3 - 4 553 111
          AK150805.1 - 3 - 4 553 111
          AK151351.1 - 3 - 4 547 111
          AK151546.1 - 3 - 4 553 111
          AK152141.1 - 3 - 4 553 111
          AK152217.1 - 3 - 4 553 111
          AK152791.1 - 3 - 4 553 111
          AK152834.1 - 3 - 4 553 111
          AK153365.1 - 3 - 4 553 111
          AK159606.1 - 3 - 4 553 111
          AK159729.1 - 3 - 4 553 111
          AK159965.1 - 3 - 4 553 111
          AK161537.1 - 3 - 4 553 111
          AK167796.1 - 3 - 4 553 111
          AK168330.1 - 3 - 4 551 111
          AK168336.1 - 3 - 4 535 111
          AK168612.1 - 3 - 4 553 111
          AK168893.1 - 3 - 4 553 111
          AK169265.1 - 3 - 4 550 111
          AK170603.1 - 3 - 4 550 111
          BC008549.1 - 3 - 4 541 3591404 111
          BC093512.1 - 3 - 4 532 5124376 111
          J05185.1 - 3 - 4 615 111
          X06453.1 - 3 - 4 555 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004215.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene P4hb

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          peptidyl-proline hydroxylation to 4-hydroxy-L-proline
          response to endoplasmic reticulum stress
          cell redox homeostasis
          cellular response to hypoxia
          regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          RNA binding
          protein disulfide isomerase activity
          procollagen-proline 4-dioxygenase activity
          isomerase activity
          enzyme binding
          peptidyl-proline 4-dioxygenase activity
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100040209.mm.1 TC1100004215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040209.mm.1 NM_011032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040209.mm.1 NM_011032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040215.mm.1 TC1100004215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040215.mm.1 NM_011032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040215.mm.1 NM_011032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040210.mm.1 TC1100004215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040210.mm.1 NM_011032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040210.mm.1 NM_011032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040212.mm.1 TC1100004215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040212.mm.1 NM_011032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040212.mm.1 NM_011032.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040211.mm.1 TC1100004215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040211.mm.1 NM_011032.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040211.mm.1 NM_011032.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 333587 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-85f78c8899-kf4dt:80/100.66.130.62:80.
          git-commit: 7f1fbef35c8161921da6e3861d81379c0ca55d8f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix