Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01243785_g1
          See other RBCK1 GE Assays ›
          Gene Symbol:
          Rbck1
          Gene Name:
          RanBP-type and C3HC4-type zinc finger containing 1
          Gene Aliases:
          AL033326, HOIL-1, HOIL-1L, UIP28, Ubce7ip3
          Chromosome Location:
          Chr.2: 152316334 - 152332643 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01243785_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbck1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001083921.1 NP_001077390.1 9 - 10 1577 66
          NM_019705.3 NP_062679.2 10 - 11 1418 66
          GenBank mRNA AF124663.1 - 8 - 9 1187 66
          AK085559.1 - 9 - 10 1566 66
          AK146176.1 - 10 - 11 1592 66
          AK166075.1 - 9 - 10 1545 66
          AK186100.1 - 1 - 2 190 66
          AK199876.1 - 2 - 3 215 66
          AK219095.1 - 3 - 4 343 66
          BC032089.1 - 2 - 3 1022 66
          BC034555.1 - 10 - 11 1414 4192310 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004894.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbck1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          negative regulation of NF-kappaB transcription factor activity
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of transcription by RNA polymerase II
          T cell receptor signaling pathway
          positive regulation of NF-kappaB transcription factor activity
          negative regulation of necroptotic process
          protein linear polyubiquitination
          positive regulation of NIK/NF-kappaB signaling
          positive regulation of extrinsic apoptotic signaling pathway
          double-stranded DNA binding
          ubiquitin-protein transferase activity
          protein kinase C binding
          protein binding
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          ubiquitin binding
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200041552.mm.1 TC0200004894.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041552.mm.1 NM_001083921.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041552.mm.1 NM_001083921.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041552.mm.1 NM_019705.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041552.mm.1 NM_019705.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041553.mm.1 TC0200004894.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041553.mm.1 NM_001083921.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041553.mm.1 NM_001083921.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041553.mm.1 NM_019705.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041553.mm.1 NM_019705.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0