Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01243844_g1
          See other RAD21L GE Assays ›
          Gene Symbol:
          Rad21l
          Gene Name:
          RAD21-like (S. pombe)
          Gene Aliases:
          Gm14160, MeiRAD21L, Rad21l1
          Chromosome Location:
          Chr.2: 151644366 - 151670622 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01243844_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rad21l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001276400.1 NP_001263329.1 8 - 9 1081 105
          GenBank mRNA AB574185.1 - 8 - 9 1081 105
          AB602048.1 - 8 - 9 1081 105
          HQ603828.1 - 8 - 9 1081 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004879.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          double-strand break repair
          chromosome segregation
          homologous chromosome pairing at meiosis
          synaptonemal complex assembly
          spermatogenesis
          fertilization
          meiotic cell cycle
          meiotic attachment of telomere to nuclear envelope
          seminiferous tubule development
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200041467.mm.1 TC0200004879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041467.mm.1 NM_001276400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041468.mm.1 TC0200004879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041468.mm.1 NM_001276400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041471.mm.1 TC0200004879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041471.mm.1 NM_001276400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041472.mm.1 TC0200004879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041472.mm.1 NM_001276400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041474.mm.1 TC0200004879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041474.mm.1 NM_001276400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041475.mm.1 TC0200004879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041475.mm.1 NM_001276400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041466.mm.1 TC0200004879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041466.mm.1 NM_001276400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041473.mm.1 TC0200004879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041473.mm.1 NM_001276400.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041470.mm.1 TC0200004879.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041470.mm.1 NM_001276400.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0