Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01245874_g1
          See other RPL27 GE Assays ›
          Gene Symbol:
          Rpl27
          Gene Name:
          ribosomal protein L27
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 101442245 - 101445596 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01245874_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011289.3 NP_035419.1 2 - 3 291 57
          GenBank mRNA AF214527.1 - 2 - 3 134 57
          AK002945.1 - 2 - 3 108 57
          AK008120.1 - 2 - 3 116 57
          AK010627.1 - 2 - 3 109 57
          AK012566.1 - 2 - 3 111 57
          AK088211.1 - 2 - 3 100 57
          AK132027.1 - 2 - 3 119 57
          AK146291.1 - 2 - 3 119 57
          BC024366.1 - 2 - 3 104 4986323 57
          BC082284.1 - 2 - 3 104 5339848 57
          BC090395.1 - 2 - 3 95 5062830 57
          BQ748836.1 - 2 - 3 96 57
          CN831363.1 - 295 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001656.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl27

          Gene Ontology Categories:

          Process(es)

          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073901 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073902 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048684 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956948 BQ748836.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956948 AK088211.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956948 AF214527.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956948 AK002945.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956948 AK008120.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971440 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138937 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163373 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956948 BQ748836.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956948 AK088211.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956948 AF214527.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956948 AK002945.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956948 AK008120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048684 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P168738 BQ748836.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P168738 AK088211.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P168738 AF214527.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P168738 AK002945.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P168738 AK008120.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073897 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163373 - -
          Whole Mouse Genome G4122A A_51_P168738 BQ748836.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P168738 AK088211.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P168738 AF214527.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P168738 AK002945.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P168738 AK008120.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110684 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112097 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163373 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09394022 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09366484 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09366487 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09366506 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073901 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28073902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08994868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08994865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09565785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09209199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09180786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08994879 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381405 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381407 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381403 - -
          Affymetrix Mouse Gene 1.0 ST Array 10453691 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10360129 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389876 - -
          Affymetrix Mouse Gene 1.1 ST Array 10453690 - -
          Affymetrix Mouse Gene 1.1 ST Array 10389875 - -
          Affymetrix Mouse Gene 1.1 ST Array 10381402 - -
          Affymetrix Mouse Gene 1.1 ST Array 10360128 - -
          Mouse Genome 430 2.0 Array 1453096_PM_x_at - -
          Mouse Genome 430 2.0 Array 1453096_x_at - -
          Mouse Genome 430 2.0 Array 1416099_at - -
          Mouse Genome 430 2.0 Array 1416099_PM_at - -
          Mouse Genome 430 2.0 Array 1448217_PM_a_at - -
          Mouse Genome 430 2.0 Array 1448217_a_at - -
          Affymetrix Mouse Clariom S Array PSR1100015518.mm.1 TC1100001656.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015518.mm.1 NM_011289.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015518.mm.1 NM_011289.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015525.mm.1 TC1100001656.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015525.mm.1 NM_011289.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015525.mm.1 NM_011289.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015510.mm.1 TC1100001656.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015510.mm.1 NM_011289.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015510.mm.1 NM_011289.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015516.mm.1 TC1100001656.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015516.mm.1 NM_011289.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015516.mm.1 NM_011289.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015523.mm.1 TC1100001656.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015523.mm.1 NM_011289.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015523.mm.1 NM_011289.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015509.mm.1 TC1100001656.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015509.mm.1 NM_011289.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015509.mm.1 NM_011289.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015517.mm.1 TC1100001656.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015517.mm.1 NM_011289.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015517.mm.1 NM_011289.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015522.mm.1 TC1100001656.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015522.mm.1 NM_011289.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015522.mm.1 NM_011289.3 Refseq
          Mouse Expression Set 430 1416099_at - -
          Mouse Expression Set 430 1448217_PM_a_at - -
          Mouse Expression Set 430 1453096_PM_x_at - -
          Mouse Expression Set 430 1453096_x_at - -
          Mouse Expression Set 430 1448217_a_at - -
          Mouse Expression Set 430 1416099_PM_at - -
          Mouse Genome 430A 2.0 Array 1448217_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1416099_at - -
          Mouse Genome 430A 2.0 Array 1453096_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1448217_a_at - -
          Mouse Genome 430A 2.0 Array 1453096_x_at - -
          Mouse Genome 430A 2.0 Array 1416099_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 893586 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1940088 - -
          Illumina mouse-6 v1.0 expression beadchip 1940088 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001940088 - -
          mouse-6 v1.1 (Illumina) scl019942.4_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 1940088 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl019942.4_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1940088 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2999797 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2748070 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2999797 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl019942.4_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0