Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01246598_m1
          See other MIR22HG GE Assays ›
          Gene Symbol:
          Mir22hg
          Gene Name:
          Mir22 host gene (non-protein coding)
          Gene Aliases:
          2210403K04Rik, mir-22
          Chromosome Location:
          Chr.11: 75461539 - 75466690 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01246598_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mir22hg
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030711.1
          1 - 2 216 98
          GenBank mRNA AK008813.1 - LIT3441
          2210403K04Rik
          1 - 2 108 98
          AK018628.1 -
          1 - 2 126 98
          AK021103.1 -
          1 - 2 217 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BE625629.1 - 1 - 2 40 Chr. 11 - 75461715 - 75469663 [+] on Build GRCm38 98
          GenBankmRNA BI100916.1 - 1 - 2 82 Chr. 11 - 75461673 - 75465721 [+] on Build GRCm38 98
          GenBankmRNA CB953026.1 - 1 - 2 74 Chr. 11 - 75461682 - 75465808 [+] on Build GRCm38 98
          GenBankmRNA AA068787.1 - 1 - 2 Chr. 11 - 75461692 - 75465501 [+] on Build GRCm38 98
          GenBankmRNA BY720809.1 - 1 - 2 122 Chr. 11 - 75461633 - 75465672 [+] on Build GRCm38 98
          GenBankmRNA BY340600.1 - 1 - 2 112 Chr. 11 - 75461644 - 75463932 [+] on Build GRCm38 98
          GenBankmRNA BY708710.1 - 111 Chr. - - [-] on Build 98
          GenBankmRNA BI792530.1 - 1 - 2 108 Chr. 11 - 75461647 - 75465616 [+] on Build GRCm38 98
          GenBankmRNA BE309708.1 - 1 - 2 53 Chr. 11 - 75461702 - 75465632 [+] on Build GRCm38 98
          GenBankmRNA BG296866.1 - 1 - 2 93 Chr. 11 - 75461662 - 75465729 [+] on Build GRCm38 98
          GenBankmRNA CF585631.1 - 187 Chr. - - [-] on Build 98
          GenBankmRNA BY728217.1 - 1 - 2 210 Chr. 11 - 75461545 - 75465580 [+] on Build GRCm38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream WD repeat domain 81 Wdr81 BC054822,
          Gm883,
          mFLJ00182
          -6.9kb Anti-sense Chr. 11 - 75440943 - 75454717 on NCBI Build GRCm38
          Downstream TLC domain containing 2 Tlcd2 4.2kb Sense Chr. 11 - 75468024 - 75470899 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004269.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mus musculus Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100040864.mm.1 TC1100004269.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100040864.mm.1 NR_030711.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100040864.mm.1 NR_030711.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009623.mm.1 TC1100004269.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009623.mm.1 NR_029739.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009623.mm.1 NR_029739.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009623.mm.1 NR_030711.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009623.mm.1 NR_030711.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009619.mm.1 TC1100004269.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009619.mm.1 NR_030711.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009619.mm.1 NR_030711.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0