Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01246600_m1
          See other MIR22HG GE Assays ›
          Gene Symbol:
          Mir22hg
          Gene Name:
          Mir22 host gene (non-protein coding)
          Gene Aliases:
          2210403K04Rik, mir-22
          Chromosome Location:
          Chr.11: 75461539 - 75466690 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01246600_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mir22hg
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030711.1
          2 - 3 387 68
          GenBank mRNA AK008813.1 - LIT3441
          2210403K04Rik
          2 - 3 279 68
          AK018628.1 -
          2 - 3 297 68
          AK021103.1 -
          2 - 3 388 68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BI100916.1 - 2 - 3 253 Chr. 11 - 75461673 - 75465721 [+] on Build GRCm38 68
          GenBankmRNA CB953026.1 - 2 - 3 245 Chr. 11 - 75461682 - 75465808 [+] on Build GRCm38 68
          GenBankmRNA AA068787.1 - 2 - 3 234 Chr. 11 - 75461692 - 75465501 [+] on Build GRCm38 68
          GenBankmRNA CA578044.1 - 1 - 2 162 Chr. 11 - 75463703 - 75465719 [+] on Build GRCm38 68
          GenBankmRNA BF786183.1 - 48 Chr. - - [-] on Build 68
          GenBankmRNA BE380224.1 - 2 - 3 201 Chr. 11 - 75461725 - 75465724 [+] on Build GRCm38 68
          GenBankmRNA AA823029.1 - 2 - 3 251 Chr. 11 - 75461682 - 75465553 [+] on Build GRCm38 68
          GenBankmRNA BY720809.1 - 2 - 3 293 Chr. 11 - 75461633 - 75465672 [+] on Build GRCm38 68
          GenBankmRNA AA467118.1 - 1 - 2 118 Chr. 11 - 75463745 - 75465518 [+] on Build GRCm38 68
          GenBankmRNA BU847963.1 - 270 Chr. - - [-] on Build 68
          GenBankmRNA BY708710.1 - 282 Chr. - - [-] on Build 68
          GenBankmRNA BI792530.1 - 2 - 3 279 Chr. 11 - 75461647 - 75465616 [+] on Build GRCm38 68
          GenBankmRNA BE309708.1 - 2 - 3 224 Chr. 11 - 75461702 - 75465632 [+] on Build GRCm38 68
          GenBankmRNA BG296866.1 - 2 - 3 264 Chr. 11 - 75461662 - 75465729 [+] on Build GRCm38 68
          GenBankmRNA AA711845.1 - 2 - 3 187 Chr. 11 - 75461735 - 75465647 [+] on Build GRCm38 68
          GenBankmRNA CF585631.1 - 358 Chr. - - [-] on Build 68
          GenBankmRNA BY728217.1 - 2 - 3 381 Chr. 11 - 75461545 - 75465580 [+] on Build GRCm38 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream WD repeat domain 81 Wdr81 BC054822,
          Gm883,
          mFLJ00182
          -9.0kb Anti-sense Chr. 11 - 75440943 - 75454717 on NCBI Build GRCm38
          Downstream TLC domain containing 2 Tlcd2 2.3kb Sense Chr. 11 - 75468024 - 75470899 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004269.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007738 AK008813.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007738 AK018628.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007738 AK021103.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2007738 NR_030711.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779985 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323528 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007738 AK008813.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007738 AK018628.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007738 AK021103.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2007738 NR_030711.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07400298 - -
          Whole Mouse Genome G4122A A_51_P144801 AK008813.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144801 AK018628.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144801 AW124019.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144801 NR_030711.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125243 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07400298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1428562_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10378568 - -
          Mouse Genome 430 2.0 Array 1428562_at - -
          Mouse Genome 430 2.0 Array 1428562_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100009624.mm.1 TC1100004269.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009624.mm.1 NR_030711.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009624.mm.1 NR_030711.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009626.mm.1 TC1100004269.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100009626.mm.1 NR_030711.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009626.mm.1 NR_030711.1 Refseq
          Mouse Expression Set 430 1428562_PM_at - -
          Mouse Expression Set 430 1428562_at - -
          Affymetrix HT Mouse Genome 430B Array 1428562_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428562_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0