Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01247543_m1
          See other IST1 GE Assays ›
          Gene Symbol:
          Ist1
          Gene Name:
          increased sodium tolerance 1 homolog (yeast)
          Gene Aliases:
          2400003C14Rik, AW536298, mKIAA0174
          Chromosome Location:
          Chr.8: 109671321 - 109693294 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01247543_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ist1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028018.2 NP_082294.1 1 - 2 109 65
          GenBank mRNA AK010262.1 - 1 - 2 79 65
          AK050468.1 - 1 - 2 76 65
          AK088192.1 - 1 - 2 64 65
          AK152830.1 - 1 - 2 76 65
          AK161899.1 - 1 - 2 65 65
          BC017605.1 - 1 - 2 75 4507347 65
          BC039052.1 - 1 - 2 52 2631157 65
          BY160969.1 - 1 - 2 110 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002908.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK010262.1, AK050468.1, AK088192.1, AK152830.1, BC017605.1, BC039052.1, NM_028018.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ist1

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokinesis
          positive regulation of defense response to virus by host
          cell cycle
          protein localization
          abscission
          viral release from host cell
          establishment of protein localization
          positive regulation of proteolysis
          viral capsid secondary envelopment
          positive regulation of collateral sprouting
          cell division
          positive regulation of mitophagy in response to mitochondrial depolarization
          protein binding
          protein domain specific binding
          protein-containing complex binding
          MIT domain binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800022380.mm.1 TC0800002908.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800022380.mm.1 NM_028018.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022380.mm.1 NM_028018.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022381.mm.1 TC0800002908.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800022381.mm.1 NM_028018.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022381.mm.1 NM_028018.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022383.mm.1 TC0800002908.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800022383.mm.1 NM_028018.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022383.mm.1 NM_028018.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 914262 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0