Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01248031_m1
          See other SULF2 GE Assays ›
          Gene Symbol:
          Sulf2
          Gene Name:
          sulfatase 2
          Gene Aliases:
          2010004N24Rik, AU020235, MSulf-2, mKIAA1247
          Chromosome Location:
          Chr.2: 166073899 - 166155683 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01248031_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sulf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252578.1 NP_001239507.1 6 - 7 1318 67
          NM_001252579.1 NP_001239508.1 6 - 7 1164 67
          NM_028072.5 NP_082348.2 6 - 7 1160 67
          GenBank mRNA AK034712.1 - 6 - 7 1145 67
          AK036685.1 - 6 - 7 1096 67
          AK049170.1 - 1 - 2 75 67
          AK081643.1 - 6 - 7 1118 67
          AK129316.1 - 6 - 7 1318 67
          AK133336.1 - 1134 67
          AK165183.1 - 6 - 7 1072 67
          AK216662.1 - 3 - 4 382 67
          AY101177.1 - 5 - 6 979 67
          BC062900.1 - 6 - 7 1117 67
          BC141086.1 - 5 - 6 913 9055698 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005164.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sulf2

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          chondrocyte development
          glomerular filtration
          sulfur compound metabolic process
          metabolic process
          positive regulation of vascular endothelial growth factor production
          esophagus smooth muscle contraction
          positive regulation of Wnt signaling pathway
          heparan sulfate proteoglycan metabolic process
          glomerular basement membrane development
          glial cell-derived neurotrophic factor receptor signaling pathway
          negative regulation of fibroblast growth factor receptor signaling pathway
          embryonic skeletal system development
          cartilage development
          bone development
          innervation
          catalytic activity
          arylsulfatase activity
          calcium ion binding
          N-acetylglucosamine-6-sulfatase activity
          sulfuric ester hydrolase activity
          hydrolase activity
          alkyl sulfatase activity
          endosulfan hemisulfate sulfatase activity
          endosulfan sulfate hydrolase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200044160.mm.1 TC0200005164.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044160.mm.1 NM_001252578.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044160.mm.1 NM_001252578.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044160.mm.1 NM_001252579.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044160.mm.1 NM_001252579.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044160.mm.1 NM_028072.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044160.mm.1 NM_028072.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044159.mm.1 TC0200005164.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200044159.mm.1 NM_001252578.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044159.mm.1 NM_001252578.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044159.mm.1 NM_001252579.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044159.mm.1 NM_001252579.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044159.mm.1 NM_028072.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200044159.mm.1 NM_028072.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0