Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01249390_g1
          See other MRPL12 GE Assays ›
          Gene Symbol:
          Mrpl12
          Gene Name:
          mitochondrial ribosomal protein L12
          Gene Aliases:
          0610034O11Rik, 1500031N16Rik, MRP-L12, Rpml12
          Chromosome Location:
          Chr.11: 120484669 - 120488754 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01249390_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027204.2 NP_081480.2 4 - 5 510 111
          GenBank mRNA AK002757.1 - 4 - 5 520 111
          AK005328.1 - 4 - 5 541 111
          AK028168.1 - 4 - 5 521 111
          AK134816.1 - 4 - 5 543 111
          AK166969.1 - 4 - 5 539 111
          AK168337.1 - 4 - 5 513 111
          BC005712.1 - 4 - 5 510 3582724 111
          BC019565.1 - 4 - 5 508 4219731 111

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrpl12

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial transcription
          translation
          positive regulation of transcription, DNA-templated
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184855 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184858 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184856 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143142 AK166969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143142 AK168337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143142 AK134816.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143142 BC005712.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143142 BC019565.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143142 AK166969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143142 AK168337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143142 AK134816.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143142 BC005712.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143142 BC019565.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876390 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153027 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135293 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143142 AK166969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143142 AK168337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143142 AK134816.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143142 BC005712.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143142 BC019565.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143142 AK166969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143142 AK168337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143142 AK134816.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143142 BC005712.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143142 BC019565.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P143142 AK166969.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P143142 AK168337.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P143142 AK134816.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P143142 BC005712.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P143142 BC019565.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P143142 AK166969.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P143142 AK168337.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P143142 AK134816.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P143142 BC005712.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P143142 BC019565.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28184880 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135293 - -
          Whole Mouse Genome G4122A A_51_P143142 AK166969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143142 AK168337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143142 AK134816.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143142 BC005712.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143142 BC019565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143142 AK166969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143142 AK168337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143142 AK134816.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143142 BC005712.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143142 BC019565.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140685 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117773 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135293 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09186888 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09186891 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09186861 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09186876 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184856 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184858 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28184855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28184916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09585948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09244959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09585949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09186876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09186891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09186889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09244960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28184870 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10535371 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383392 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383391 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383393 - -
          Affymetrix HT MG-430 PM Array Plate 1452048_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10383389 - -
          Affymetrix Mouse Gene 1.1 ST Array 10535369 - -
          Mouse Genome 430 2.0 Array 1452048_PM_at - -
          Mouse Genome 430 2.0 Array 1452048_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56282_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56282_PM_at - -
          Murine Genome U74v2 Set 160431_PM_at - -
          Murine Genome U74v2 Set 160431_at - -
          Affymetrix HT Mouse Genome 430A Array 1452048_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452048_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3120112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2735335 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2735335 - -
          mouse-6 v1.1 (Illumina) scl056282.5_222-S - -
          Illumina Sentrix mouse-6.v1 bead array 3120112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056282.5_222-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2883147 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2883147 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl056282.5_222-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0