Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01249513_g1
          See other 9330175E14RIK GE Assays ›
          Gene Symbol:
          9330175E14Rik
          Gene Name:
          RIKEN cDNA 9330175E14 gene
          Gene Aliases:
          A630085K21
          Chromosome Location:
          Chr.8: 94433127 - 94435103 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01249513_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9330175E14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015514.2 2 - 3 518 61
          GenBank mRNA AK034303.1 - 60 61
          AK053303.1 - 2 - 3 520 61

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK034303.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2276224 BC103768.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2276224 AK034303.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 621366 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132712 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2276224 BC103768.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2276224 AK034303.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417945 BC103768.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P417945 AK034303.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P311976 NR_015514.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P311976 AK034303.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P311976 AK053303.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07388951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07405446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07427840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06916303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07388952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07365298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07388950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07405448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26077495 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10580755 - -
          Affymetrix HT MG-430 PM Array Plate 1446547_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1460134_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10580754 - -
          Mouse Genome 430 2.0 Array 1446547_PM_at - -
          Mouse Genome 430 2.0 Array 1460134_at - -
          Mouse Genome 430 2.0 Array 1446547_at - -
          Mouse Genome 430 2.0 Array 1460134_PM_at - -
          Mouse Expression Set 430 1460134_PM_at - -
          Mouse Expression Set 430 1446547_at - -
          Mouse Expression Set 430 1460134_at - -
          Mouse Expression Set 430 1446547_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446547_at - -
          Affymetrix HT Mouse Genome 430B Array 1446547_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460134_at - -
          Affymetrix HT Mouse Genome 430B Array 1460134_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0