Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029282.1 | NP_083558.1 | 1 - 2 | 2348 | 78 | |
GenBank mRNA | AF332067.1 | - | 1 - 2 | 2331 | 78 | |
AF332068.1 | - | 1 - 2 | 2331 | 78 | ||
AK128940.1 | - | 2 - 3 | 2398 | 78 | ||
AK195103.1 | - | 1 - 2 | 81 | 78 | ||
AK219901.1 | - | 1 - 2 | 81 | 78 | ||
BC053067.1 | - | 1 - 2 | 2348 | 5698252 | 78 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |