Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01250461_g1
          See other ARRB2 GE Assays ›
          Gene Symbol:
          Arrb2
          Gene Name:
          arrestin, beta 2
          Gene Aliases:
          AI326910, AW122872
          Chromosome Location:
          Chr.11: 70432580 - 70440828 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01250461_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arrb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271358.1 NP_001258287.1 12 - 13 1156 74
          NM_001271359.1 NP_001258288.1 12 - 13 1156 74
          NM_001271360.1 NP_001258289.1 12 - 13 1156 74
          NM_145429.5 NP_663404.1 12 - 13 1156 74
          GenBank mRNA AK089542.1 - 12 - 13 1092 74
          AK154874.1 - 12 - 13 1076 74
          AK159317.1 - 12 - 13 1084 74
          AK162889.1 - 12 - 13 1093 74
          AK170647.1 - 12 - 13 1103 74
          AK170889.1 - 12 - 13 1066 74
          BC016642.1 - 12 - 13 1038 2651372 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000950.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arrb2

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          G protein-coupled receptor internalization
          desensitization of G protein-coupled receptor signaling pathway by arrestin
          positive regulation of receptor internalization
          transcription by RNA polymerase II
          transport
          endocytosis
          signal transduction
          transforming growth factor beta receptor signaling pathway
          G protein-coupled receptor signaling pathway
          adult walking behavior
          regulation of G protein-coupled receptor signaling pathway
          negative regulation of signal transduction
          protein transport
          protein ubiquitination
          negative regulation of protein ubiquitination
          positive regulation of protein ubiquitination
          receptor internalization
          negative regulation of NF-kappaB transcription factor activity
          positive regulation of synaptic transmission, dopaminergic
          negative regulation of interleukin-1 beta production
          negative regulation of interleukin-12 production
          negative regulation of interleukin-6 production
          negative regulation of tumor necrosis factor production
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of toll-like receptor signaling pathway
          negative regulation of GTPase activity
          negative regulation of smooth muscle cell apoptotic process
          follicle-stimulating hormone signaling pathway
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of natural killer cell mediated cytotoxicity
          positive regulation of peptidyl-tyrosine phosphorylation
          detection of temperature stimulus involved in sensory perception of pain
          positive regulation of protein kinase B signaling
          positive regulation of calcium ion transport
          cell chemotaxis
          regulation of androgen receptor signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          negative regulation of release of cytochrome c from mitochondria
          positive regulation of DNA biosynthetic process
          G protein-coupled receptor binding
          signaling receptor binding
          protein binding
          enzyme binding
          protein domain specific binding
          ubiquitin protein ligase binding
          alpha-1A adrenergic receptor binding
          alpha-1B adrenergic receptor binding
          angiotensin receptor binding
          type 1 angiotensin receptor binding
          D1 dopamine receptor binding
          follicle-stimulating hormone receptor binding
          type 2A serotonin receptor binding
          platelet activating factor receptor binding
          protein kinase B binding
          protein-containing complex binding
          mitogen-activated protein kinase binding
          molecular adaptor activity
          14-3-3 protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100008342.mm.1 TC1100000950.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008342.mm.1 NM_001271358.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008342.mm.1 NM_001271359.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008342.mm.1 NM_001271360.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008342.mm.1 NM_145429.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008344.mm.1 TC1100000950.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008344.mm.1 NM_001271358.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008344.mm.1 NM_001271359.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008344.mm.1 NM_001271360.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008344.mm.1 NM_145429.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008345.mm.1 TC1100000950.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100008345.mm.1 NM_001271358.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100008345.mm.1 NM_001271359.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0