Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01251779_m1
          See other PABPC1L GE Assays ›
          Gene Symbol:
          Pabpc1l
          Gene Name:
          poly(A) binding protein, cytoplasmic 1-like
          Gene Aliases:
          1810053B01Rik, Epab
          Chromosome Location:
          Chr.2: 164024770 - 164050538 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01251779_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pabpc1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001114079.2 NP_001107551.1 8 - 9 1484 75
          GenBank mRNA BC158030.1 - 8 - 9 1484 9007333 75
          EU669908.1 - 8 - 9 1227 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002500.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          oocyte maturation
          mRNA polyadenylation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P08881 AK007853.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P08881 NM_001114079.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 114630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634421 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 680839 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 588578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270974 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 258122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 337802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 958345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 588928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448488 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 708437 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25727 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 53677 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P08881 AK007853.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P08881 NM_001114079.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21815884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01160472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01596126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01979987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01914654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02237781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01806514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01231811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01532217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01942977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02237785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01413901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01551011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01823455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01816577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01914641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01375386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01590906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02255059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01914643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01671531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01939210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01231813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01549047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01806512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01231805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01348546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01348537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01939204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01551013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01914659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02016296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01348538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01914662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02255062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21815706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02237789 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10478434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478444 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478443 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478435 - -
          Affymetrix HT MG-430 PM Array Plate 1439913_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10478432 - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381404_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 381404_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200020423.mm.1 TC0200002500.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020423.mm.1 NM_001114079.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020419.mm.1 TC0200002500.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020419.mm.1 NM_001114079.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020421.mm.1 TC0200002500.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020421.mm.1 NM_001114079.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020426.mm.1 TC0200002500.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020426.mm.1 NM_001114079.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020418.mm.1 TC0200002500.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020418.mm.1 NM_001114079.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020417.mm.1 TC0200002500.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020417.mm.1 NM_001114079.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020424.mm.1 TC0200002500.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020424.mm.1 NM_001114079.2 Refseq
          Affymetrix HT Mouse Genome 430B Array 1439913_at - -
          Affymetrix HT Mouse Genome 430B Array 1439913_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 415407 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0